L-Ascorbic acid sodium salt, 99%
L-Ascorbic acid sodium salt, 99%
Thermo Scientific Chemicals

L-Ascorbic acid sodium salt, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 352685000
Price (USD)/ Each
143.65
Online exclusive
159.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
143.65
Online exclusive
159.00 
Save 15.35 (10%)
Add to cart
L-Ascorbic acid sodium salt, 99%
Catalog number352685000
Price (USD)/ Each
143.65
Online exclusive
159.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS134-03-2
IUPAC Namesodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
Molecular FormulaC6H7NaO6
InChI KeyIFVCRSPJFHGFCG-HXPAKLQESA-N
SMILES[Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration Iodimetric98.5 to 101.5 %
Solubility(5% in water) Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only