4-(Bromomethyl)-7-methoxycoumarin, 97%, Thermo Scientific Chemicals
4-(Bromomethyl)-7-methoxycoumarin, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Bromomethyl)-7-methoxycoumarin, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 365190010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS35231-44-8
IUPAC Name4-(bromomethyl)-7-methoxy-2H-chromen-2-one
Molecular FormulaC11H9BrO3
InChI KeyCTENSLORRMFPDH-UHFFFAOYSA-N
SMILESCOC1=CC=C2C(CBr)=CC(=O)OC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to yellow to beige
Appearance (Form)Powder and/or chunks
Infrared spectrumConforms
HPLC>=96.0 %
Melting point212°C to 217°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only