3740WHI Full specifications
Case of 480 White V-Bottom ScrewTop Tubes Bulk, 480 Tubes Sterile & PCR Ready - Product Type Screw Top Tube Description V-Bottom ScrewTop Tubes Closure Color White Autoclavable No Capacity (Metric) 1.0 mL Bottom Shape V-bottom Material Polypropylene (Medical Grade) Packaging Bulk, 480 Tubes Well Shape V-bottom Barcode Non-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction Product Line Matrix™ Sterility Sterile Closure Included Yes Closure Material Virgin Class VI Medical Grade Polypropylene with Molded-in thermoplastic elastomer gasket Capacity (English) 0.0338 fl oz. Dnase Free Yes Fit for Purpose Sterile & PCR Ready Unit Size Case of 480
3740 Full specifications
- Colorless V-Bottom ScrewTop Tubes Bulk, 480 Tubes Sterile & PCR Ready Request A Quote - - Product Type Screw Top Tube Description V-Bottom ScrewTop Tubes Closure Color Colorless Autoclavable Autoclavable (with cap loosened) Capacity (Metric) 1.0 mL Bottom Shape V-bottom Material Polypropylene (Medical Grade) Packaging Bulk, 480 Tubes Well Shape V-bottom Barcode Non-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction Product Line Matrix™ Sterility Sterile Closure Material Virgin Class VI Medical Grade Polypropylene with Molded-in thermoplastic elastomer gasket Capacity (English) 0.0338 fl oz. Dnase Free Yes Closure Type Colorless caps Fit for Purpose Sterile & PCR Ready Unit Size -
3740TS also known as 3740
Full specifications
Case of 480 Colorless V-Bottom ScrewTop Tubes Case - Description V-Bottom ScrewTop Tubes Closure Color Colorless Autoclavable Autoclavable (with cap loosened) Capacity (Metric) 1.0 mL Material Virgin Class VI Medical Grade Polypropylene Package Configuration Bulk, 480/case Packaging Case Well Shape V-bottom Barcode Non-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction Product Line Matrix™ Sterility Sterile Unit Size Case of 480
3740RED Full specifications
Case of 480 Red V-Bottom ScrewTop Tubes Bulk, 480 Tubes Sterile & PCR Ready - Product Type Screw Top Tube Description V-Bottom ScrewTop Tubes Closure Color Red Autoclavable No Capacity (Metric) 1.0 mL Bottom Shape V-bottom Material Polypropylene (Medical Grade) Packaging Bulk, 480 Tubes Well Shape V-bottom Barcode Non-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction Product Line Matrix™ Sterility Sterile Closure Included Yes Closure Material Virgin Class VI Medical Grade Polypropylene with Molded-in thermoplastic elastomer gasket Capacity (English) 0.0338 fl oz. Dnase Free Yes Fit for Purpose Sterile & PCR Ready Unit Size Case of 480
3740YEL Full specifications
Case of 480 Yellow V-Bottom ScrewTop Tubes Bulk, 480 Tubes Sterile & PCR Ready - Product Type Screw Top Tube Description V-Bottom ScrewTop Tubes Closure Color Yellow Autoclavable No Capacity (Metric) 1.0 mL Bottom Shape V-bottom Material Polypropylene (Medical Grade) Packaging Bulk, 480 Tubes Well Shape V-bottom Barcode Non-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction Product Line Matrix™ Sterility Sterile Closure Included Yes Closure Material Virgin Class VI Medical Grade Polypropylene with Molded-in thermoplastic elastomer gasket Capacity (English) 0.0338 fl oz. Dnase Free Yes Fit for Purpose Sterile & PCR Ready Unit Size Case of 480
3740BLU Full specifications
Case of 480 Blue V-Bottom ScrewTop Tubes Bulk, 480 Tubes Sterile & PCR Ready - Product Type Screw Top Tube Description V-Bottom ScrewTop Tubes Closure Color Blue Autoclavable No Capacity (Metric) 1.0 mL Bottom Shape V-bottom Material Polypropylene (Medical Grade) Packaging Bulk, 480 Tubes Well Shape V-bottom Barcode Non-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction Product Line Matrix™ Sterility Sterile Closure Included Yes Closure Material Virgin Class VI Medical Grade Polypropylene with Molded-in thermoplastic elastomer gasket Capacity (English) 0.0338 fl oz. Dnase Free Yes Fit for Purpose Sterile & PCR Ready Unit Size Case of 480
3740GRE Full specifications
Case of 480 Green V-Bottom ScrewTop Tubes Bulk, 480 Tubes Sterile & PCR Ready - Product Type Screw Top Tube Description V-Bottom ScrewTop Tubes Closure Color Green Autoclavable No Capacity (Metric) 1.0 mL Bottom Shape V-bottom Material Polypropylene (Medical Grade) Packaging Bulk, 480 Tubes Well Shape V-bottom Barcode Non-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction Product Line Matrix™ Sterility Sterile Closure Included Yes Closure Material Virgin Class VI Medical Grade Polypropylene with Molded-in thermoplastic elastomer gasket Capacity (English) 0.0338 fl oz. Dnase Free Yes Fit for Purpose Sterile & PCR Ready Unit Size Case of 480
3740PUR Full specifications
Case of 480 Purple V-Bottom ScrewTop Tubes Bulk, 480 Tubes Sterile & PCR Ready - Product Type Screw Top Tube Description V-Bottom ScrewTop Tubes Closure Color Purple Autoclavable No Capacity (Metric) 1.0 mL Bottom Shape V-bottom Material Polypropylene (Medical Grade) Packaging Bulk, 480 Tubes Well Shape V-bottom Barcode Non-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction Product Line Matrix™ Sterility Sterile Closure Included Yes Closure Material Virgin Class VI Medical Grade Polypropylene with Molded-in thermoplastic elastomer gasket Capacity (English) 0.0338 fl oz. Dnase Free Yes Fit for Purpose Sterile & PCR Ready Unit Size Case of 480
3741 Full specifications
Case of 480 Colorless V-Bottom ScrewTop Tubes 5 Latch Racks of 96 Tubes Sterile & PCR Ready - Product Type Screw Top Tube Description V-Bottom ScrewTop Tubes Closure Color Colorless Autoclavable No Capacity (Metric) 1.0 mL Bottom Shape V-bottom Material Polypropylene (Medical Grade) Packaging 5 Latch Racks of 96 Tubes Well Shape V-bottom Barcode Non-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction Product Line Matrix™ Sterility Sterile Closure Material Virgin Class VI Medical Grade Polypropylene with Molded-in thermoplastic elastomer gasket Capacity (English) 0.0338 fl oz. Dnase Free Yes Closure Type Colorless Cap Fit for Purpose Sterile & PCR Ready Unit Size Case of 480
3741RED Full specifications
Case of 480 Red V-Bottom ScrewTop Tubes 5 Latch Racks of 96 Tubes Sterile & PCR Ready - Product Type Screw Top Tube Description V-Bottom ScrewTop Tubes Closure Color Red Autoclavable No Capacity (Metric) 1.0 mL Bottom Shape V-bottom Material Polypropylene (Medical Grade) Packaging 5 Latch Racks of 96 Tubes Well Shape V-bottom Barcode Non-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction Product Line Matrix™ Sterility Sterile Closure Included Yes Closure Material Virgin Class VI Medical Grade Polypropylene with Molded-in thermoplastic elastomer gasket Capacity (English) 0.0338 fl oz. Dnase Free Yes Fit for Purpose Sterile & PCR Ready Unit Size Case of 480