Thermo Scientific Chemicals

4-bromo-1-fluoro-2-nitrobenzene, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): 380890050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-bromo-1-fluoro-2-nitrobenzene, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): 380890050
5 g, Glass bottle, Each
数量
パッケージ
製品番号(カタログ番号): 380890050

化学物質識別子

CAS
364-73-8
IUPAC Name
4-bromo-1-fluoro-2-nitrobenzene
Molecular Formula
C6H3BrFNO2
InChI Key
UQEANKGXXSENNF-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC(Br)=CC=C1F
Appearance (Color)
Clear yellow to brown
GC
>=97.5 %
Infrared spectrum
Conforms
Appearance (Form)
Liquid after melting
Refractive index
1.5750 to 1.5780 (20°C, 589 nm)

概要

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text