2-Methyltetrahydrofuran, 99+%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
2-Methyltetrahydrofuran, 99+%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
2-Methyltetrahydrofuran, 99+%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
2-Methyltetrahydrofuran, 99+%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
Thermo Scientific Chemicals

2-Methyltetrahydrofuran, 99+%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™

CAS: 96-47-9 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number 396630010
Price (USD)
175.65
Special offer
Online exclusive
206.00
Save 30.35 (15%)
Each
-
Add to cart
Quantity:
1 L
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
175.65
Special offer
Online exclusive
206.00
Save 30.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS96-47-9
IUPAC Name2-methyloxolane
Molecular FormulaC5H10O
InChI KeyJWUJQDFVADABEY-UHFFFAOYNA-N
SMILESCC1CCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Peroxides=<50 ppm
Stabilizer150 to 400 ppm BHT
Water=<0.002 %
GC>=99.0 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2534308Certificate of AnalysisMay 24, 2025396630010, 396631000
2525771Certificate of AnalysisFeb 07, 2025396630010, 396631000
2499421Certificate of AnalysisJan 31, 2025396630010, 396631000
2407368Certificate of AnalysisJan 30, 2025396630010, 396631000
2416215Certificate of AnalysisJan 30, 2025396630010, 396631000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection