Calcium carbonate, 98%, pure, light powder, Thermo Scientific Chemicals
Calcium carbonate, 98%, pure, light powder, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium carbonate, 98%, pure, light powder, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
View all
Plastic drum
Catalog number 403810025
Price (USD)/ Each
98.65
Online exclusive
116.00 
Save 17.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
98.65
Online exclusive
116.00 
Save 17.35 (15%)
Add to cart
Calcium carbonate, 98%, pure, light powder, Thermo Scientific Chemicals
Catalog number403810025
Price (USD)/ Each
98.65
Online exclusive
116.00 
Save 17.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namecalcium carbonate
Molecular FormulaCCaO3
InChI KeyVTYYLEPIZMXCLO-UHFFFAOYSA-L
SMILES[Ca++].[O-]C([O-])=O
Molecular Weight (g/mol)100.09
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Titration Complexometric>=97.5 %
Loss on drying=<2 % (200°C, 4 h)
Acid insoluble substances=<0.2 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only