Dinonyl phthalate, 96%, mixture of isomers
Dinonyl phthalate, 96%, mixture of isomers
Dinonyl phthalate, 96%, mixture of isomers
Dinonyl phthalate, 96%, mixture of isomers
Thermo Scientific Chemicals

Dinonyl phthalate, 96%, mixture of isomers

CAS: 84-76-4 | C26H42O4 | 418.62 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 408770250
価格(JPY)
-
見積もりを依頼する
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS84-76-4
Appearance (Form)Viscous liquid
Appearance (Color)Clear colorless to yellow-brown
Refractive index1.4820 to 1.4910 (20°C, 589 nm)
GC>=95.0 % (sum of isomers)
Infrared spectrumConforms
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Kunden, die diesen Artikel ansahen, interessierten sich auch für



ドキュメントおよびダウンロード

証明書

ロット番号Certificate TypeDateCatalog Number(s)
A0469479Certificate of Analysis2025年2月03日408770250, 408771000
A0469480Certificate of Analysis2025年1月31日408770250, 408771000
A0396781Certificate of Analysis2022年12月14日408770250, 408771000
A0397576Certificate of Analysis2022年12月14日408770250, 408771000
A0414428Certificate of Analysis2022年12月13日408770250, 408771000
5件の結果が表示されました。 上記から特定の証明書を検索します

Safety Data Sheets

こちらの製品には複数の製品番号が設定されています。

1x1 image pixel for data collection