Thermo Scientific Chemicals

New methylene Blue N, pure

Catalog number: 415530025
2.5 g, Glass bottle, Each
Thermo Scientific Chemicals

New methylene Blue N, pure

Catalog number: 415530025
2.5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 415530025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1934-16-3
IUPAC Name
3,7-bis(ethylamino)-2,8-dimethyl-5λ⁴-phenothiazin-5-ylium chloride
Molecular Formula
C18H22ClN3S
InChI Key
NZYCYASKVWSANA-UHFFFAOYSA-M
SMILES
[Cl-].CCNC1=CC2=[S+]C3=CC(NCC)=C(C)C=C3N=C2C=C1C
Appearance (Color)
Green to dark brown
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Dye content
ca 70%

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text