Calcium oxide, 97+%, for analysis, powder
Calcium oxide, 97+%, for analysis, powder
Calcium oxide, 97+%, for analysis, powder
Calcium oxide, 97+%, for analysis, powder
Thermo Scientific Chemicals

Calcium oxide, 97+%, for analysis, powder

Calcium oxide, 97+%, CaO, CAS Number-1305-78-8, gebrannter kalk, calxyl, calcia, burnt lime, calcium oxide, airlock, calcium monoxide, calx, unslaked lime, 1kg, 2.0% max., 97% min. (CaO) on dry substance (Complexometry), 2850 deg.C, CHEBI:31344, White or Yellow, 215-138-9, 56.08, Analysis | CAS: 1305-78-8 | CaO | 56.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
Packaging:
Plastic bottle
Catalog number 422830010
Price (USD)
34.65
Online Exclusive
38.00
Save 3.35 (9%)
Each
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
34.65
Online Exclusive
38.00
Save 3.35 (9%)
Each
Add to cart
Chemical Identifiers
CAS1305-78-8
IUPAC Namecalcium oxidandiide
Molecular FormulaCaO
InChI KeyBRPQOXSCLDDYGP-UHFFFAOYSA-N
SMILES[O--].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Complexometric>=97.0 % (CaO) on dry substance
Heavy metals (as Pb)=<100 ppm
Loss on ignition=<5 %
NH4OH precipitate=<2.0 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Los clientes que vieron este artículo también vieron



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471404Certificate of AnalysisMay 17, 2025422830010, 422830025
A0466692Certificate of AnalysisOct 17, 2024422830010, 422830025
A0463543Certificate of AnalysisJun 06, 2024422830010, 422830025
A0456732Certificate of AnalysisOct 12, 2023422830010, 422830025
A0452862Certificate of AnalysisMay 09, 2023422830010, 422830025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection