Benzoic acid, 99.6%, ACS reagent
Benzoic acid, 99.6%, ACS reagent
Benzoic acid, 99.6%, ACS reagent
Benzoic acid, 99.6%, ACS reagent
Thermo Scientific Chemicals

Benzoic acid, 99.6%, ACS reagent

CAS: 65-85-0 | C7H6O2 | 122.12 g/mol
数量:
25 g
500 g
2 kg
パッケージ:
Plastic bottle
製品番号(カタログ番号) 423475000
価格(JPY)
-
見積もりを依頼する
数量:
500 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS65-85-0
IUPAC Namebenzoic acid
Molecular FormulaC7H6O2
InChI KeyWPYMKLBDIGXBTP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1
さらに表示
Appearance (Color)White
Freezing point122°C to 123°C
Titration with NaOH>=99.5 %
Heavy metals (as Pb)=<5 ppm
Residue after ignition=<0.005 %
さらに表示
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

ドキュメントおよびダウンロード

証明書

ロット番号Certificate TypeDateCatalog Number(s)
A0465069Certificate of Analysis2024年8月09日423475000, 423470250, 423470020
A0462506Certificate of Analysis2024年5月13日423475000, 423470250, 423470020
A0394662Certificate of Analysis2022年12月14日423475000, 423470250, 423470020
A0408083Certificate of Analysis2022年12月13日423475000, 423470250, 423470020
A0448035Certificate of Analysis2022年12月13日423475000, 423470250, 423470020
5件の結果が表示されました。 上記から特定の証明書を検索します

Safety Data Sheets

こちらの製品には複数の製品番号が設定されています。

1x1 image pixel for data collection