N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent
N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent
N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent
N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent
Thermo Scientific™

N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent

N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, C12H14N2.2ClH, CAS Number-1465-25-4 | CAS: 1465-25-4 | C12H16Cl2N2 | 259.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number 423990050
Price (USD)
47.65
Special offer
Online exclusive
Ends: 26-Sep-2025
58.40
Save 10.75 (18%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 26-Sep-2025
58.40
Save 10.75 (18%)
Each
Add to cart
Chemical Identifiers
CAS1465-25-4
IUPAC Namedihydrogen N1-(naphthalen-1-yl)ethane-1,2-diamine dichloride
Molecular FormulaC12H16Cl2N2
InChI KeyMZNYWPRCVDMOJG-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NCCNC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
SolubilityPasses test
Appearance (Color)White to beige to pink
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
Titration Argentometric>=98.0 % (on dry substance)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0472291Certificate of AnalysisMay 17, 2025423991000, 423990050, 423990250, 423995000
A0469250Certificate of AnalysisJan 31, 2025423991000, 423990050, 423990250, 423995000
B0153644Certificate of AnalysisJul 09, 2024423991000, 423990050, 423990250, 423995000
B0156021Certificate of AnalysisJun 11, 2024423991000, 423990050, 423990250, 423995000
B0155548Certificate of AnalysisJun 11, 2024423991000, 423990050, 423990250, 423995000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link