Sodium methoxide, 0.5M solution in methanol, AcroSeal™, Thermo Scientific Chemicals
Sodium methoxide, 0.5M solution in methanol, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium methoxide, 0.5M solution in methanol, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 427221000
Price (USD)/ Each
81.65
Online exclusive
90.90 
Save 9.25 (10%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
81.65
Online exclusive
90.90 
Save 9.25 (10%)
Add to cart
Sodium methoxide, 0.5M solution in methanol, AcroSeal™, Thermo Scientific Chemicals
Catalog number427221000
Price (USD)/ Each
81.65
Online exclusive
90.90 
Save 9.25 (10%)
-
Add to cart
Chemical Identifiers
CAS124-41-4
IUPAC Namesodium methanolate
Molecular FormulaCH3NaO
InChI KeyWQDUMFSSJAZKTM-UHFFFAOYSA-N
SMILES[Na+].C[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Strength0.45 to 0.60 M
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only