Diazolidinyl urea, 95%
Diazolidinyl urea, 95%
Diazolidinyl urea, 95%
Diazolidinyl urea, 95%
Thermo Scientific Chemicals

Diazolidinyl urea, 95%

CAS: 78491-02-8 | C8H14N4O7 | 278.22 g/mol
数量:
25 g
250 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 428580250
価格(JPY)
-
見積もりを依頼する
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS78491-02-8
IUPAC Name1-[1,3-bis(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-1,3-bis(hydroxymethyl)urea
Molecular FormulaC8H14N4O7
InChI KeySOROIESOUPGGFO-UHFFFAOYNA-N
SMILESOCNC(=O)N(CO)C1N(CO)C(=O)N(CO)C1=O
さらに表示
Appearance (Color)White
Loss on drying=<3 %
Elemental Analysis19.0 to 21.5 % N
Solubility(50 mg/ml in water) Clear colorless
Appearance (Form)Powder
さらに表示
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

ドキュメントおよびダウンロード

証明書

ロット番号Certificate TypeDateCatalog Number(s)
344257Certificate of Analysis2025年1月31日428580250, 428582500
345286Certificate of Analysis2024年2月06日428580250, 428582500
343793Certificate of Analysis2022年12月14日428580250, 428582500
344046Certificate of Analysis2022年12月13日428580250, 428582500
344257Certificate of Analysis2022年7月18日428580250, 428582500
5件の結果が表示されました。 上記から特定の証明書を検索します

Safety Data Sheets

こちらの製品には複数の製品番号が設定されています。

1x1 image pixel for data collection