Applied Biosystems™

POP-7™ Polymer for 3730/3730xl DNA Analyzers

Catalog number: 4335611
Have Questions?
Applied Biosystems™

POP-7™ Polymer for 3730/3730xl DNA Analyzers

Catalog number: 4335611
Have Questions?
Catalog Number
4335611
Unit Size
30 x 28 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 x 28 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 x 28 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 x 28 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
433561130 x 28 mL
-
43639291 x 28 mL
-
43356155 x 28 mL
-
436393510 x 28 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
POP-7™ Polymer is a separation matrix for the 3730 and 3730xl DNA Analyzers. POP-7™ Polymer is customized for DNA sequencing and fragment analysis. Not suitable for use with ABI PRISM ™, 310 and 3100 series genetic analyzers. It causes syringe failure and premature array degradation. Sufficient for 8,000 - 12,000 samples.

POP-7™ Polymers for the 3730⁄3730xl Genetic Analyzers is:
• Pre-formulated, which not only saves time but also saves reproducibility by ensuring consistent formulation
• Formulated for sequencing and fragment analysis applications
• Designed to allow capillaries to be used multiple times

Reproducible Results
POP™ polymers dynamically coat the capillary wall to control electro-osmotic flow. Their definite quality and uniform consistency eliminate guesswork and help to ensure reproducibility. POP polymers are specifically formulated to separate DNA fragments of a known size range at a desired resolution and run time.

Used With
3730/3730xl Genetic Analyzer Capillary Array, 50 cm, 3730 Running Buffer, 10X, BigDye™ Terminator v1.1 Cycle Sequencing Kit, BigDye™ Terminator v3.1 Cycle Sequencing Kit, BigDye™ Terminator v3.1 Cycle Sequencing Kit, BigDye™ Terminator v3.1 Cycle Sequencing Kit, Hi-Di™ Formamide.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Accuracy
98.5% up to 900 bases (for 50 cm capillary), 98.5% up to 700 bases (for 36 cm capillary)
For Use With (Equipment)
3730 DNA Analyzer
For Use With (Application)
Standard Sequencing, Fragment Analysis (Denaturing)
On Instrument Life Usage
Stability on instrument 7 days, up to 12,000 samples/28 mL
Product Line
POP-7™
Product Type
Polymer
Quantity
30 x 28 mL
Test Time
1 hr for 36 cm capillary (std. seq.), 2 hrs for 50 cm capillary (long read seq.)

Contents & Storage

Refrigerate at 2°C to 8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text