Applied Biosystems™

POP-7™ Polymer for 3730/3730xl DNA Analyzers

Catalog number: 4363935
Have Questions?
Applied Biosystems™

POP-7™ Polymer for 3730/3730xl DNA Analyzers

Catalog number: 4363935
Have Questions?
Catalog Number
4363935
Unit Size
10 x 28 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 x 28 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 x 28 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
30 x 28 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
436393510 x 28 mL
-
43639291 x 28 mL
-
43356155 x 28 mL
-
433561130 x 28 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
POP-7™ Polymer is a separation matrix for the 3730 and 3730xl DNA Analyzers. POP-7™ Polymer is customized for DNA sequencing and fragment analysis. Not suitable for use with ABI PRISM ™, 310 and 3100 series genetic analyzers. It causes syringe failure and premature array degradation. Sufficient for 8,000 - 12,000 samples.

POP-7™ Polymers for the 3730⁄3730xl Genetic Analyzers is:
• Pre-formulated, which not only saves time but also saves reproducibility by ensuring consistent formulation
• Formulated for sequencing and fragment analysis applications
• Designed to allow capillaries to be used multiple times

Reproducible Results
POP™ polymers dynamically coat the capillary wall to control electro-osmotic flow. Their definite quality and uniform consistency eliminate guesswork and help to ensure reproducibility. POP polymers are specifically formulated to separate DNA fragments of a known size range at a desired resolution and run time.

Used With
3730/3730xl Genetic Analyzer Capillary Array, 50 cm, 3730 Running Buffer, 10X, BigDye™ Terminator v1.1 Cycle Sequencing Kit, BigDye™ Terminator v3.1 Cycle Sequencing Kit, BigDye™ Terminator v3.1 Cycle Sequencing Kit, BigDye™ Terminator v3.1 Cycle Sequencing Kit, Hi-Di™ Formamide.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Accuracy
98.5% up to 900 bases (for 50 cm capillary), 98.5% up to 700 bases (for 36 cm capillary)
For Use With (Equipment)
3730xl DNA Analyzer, 3730 DNA Analyzer
For Use With (Application)
Standard Sequencing, Fragment Analysis (Denaturing)
On Instrument Life Usage
Stability on instrument 7 days, sufficient for 8000 to 12,000 samples
Product Line
POP-7™
Product Type
Polymer
Quantity
10 x 28 mL
Test Time
1 hr for 36 cm capillary (std. seq.), 2 hrs for 50 cm capillary (long read seq.)

Contents & Storage

Store at 2°C to 8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text