Informational:Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
Informational:BioProbes™ Journal of Cell Biology Applications—Subscribe today »
Invitrogen™

Streptavidin

Catalog number: 434302
Have Questions?
Invitrogen™

Streptavidin

Catalog number: 434302
Have Questions?
Catalog Number
434302
Unit Size
5 mg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
4343025 mg
-
4343011 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Streptavidin is a bacterially derived biotin-binding protein that reportedly exhibits less nonspecific binding than avidin.

Capacity: Binding activity of streptavidin is determined to be >11 units per mg protein. One unit of binding activity is defined as that amount of protein which binds 1 μg of biotin (Green, N.M., Biochem. J., 89:599,1963)

Form: Streptavidin is provided in lyophilized form in 10 mM phosphate buffered saline, pH 7.4. Reconstitute with 1 ml of distilled or deionized water.

Storage: Store at 4°C.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Conjugate
Unconjugated
Shipping Condition
Wet Ice
Form
Lyophilized
Product Type
Streptavidin Conjugate (fluorescent)

Contents & Storage

Streptavidin is provided in lyophilized form in 10 mM phosphate buffered saline, pH 7.4. Reconstitute with 1 ml of distilled or deionized water. Store at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text