Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

Nuclease-Free Water (not DEPC-Treated)

Catalog number: 4387936
Have Questions?
Invitrogen™

Nuclease-Free Water (not DEPC-Treated)

Catalog number: 4387936
Have Questions?
Catalog Number
4387936
Unit Size
4 x 1,000 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 x 100 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 x 500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 x 50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 x 100 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 x 1,000 ml
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
43879364 x 1,000 mL
-
AM99381 x 100 mL
-
AM99301 x 500 mL
-
AM993710 x 50 mL
-
AM99395 x 100 mL
-
AM99321 x 1,000 ml
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Nuclease-free Water (not DEPC-treated) has been deionized, filtered into the final bottle, and autoclaved. It is ready to use and requires no preparation, mixing, or autoclaving. This nuclease-free water is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Using Nuclease-free Water
Because this water is not DEPC-treated, it is recommended for applications for which residual DEPC might prove detrimental (e.g., oocyte microinjection).

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Recommended Storage
Store at room temperature.
Grade
Molecular Biology
Packaging
Bottle
Treatment(s)
Not DEPC-Treated
Quantity
4 x 1000 mL
Purification Method
Autoclaved, Membrane-Filtered
Product Line
Ambion™

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text