VIC™ Dye Spectral Calibration Plate, 96-well
Product Image
Applied Biosystems™

VIC™ Dye Spectral Calibration Plate, 96-well

This VIC™ dye spectral calibration plate can be used to maintain your ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex,Read more
Have Questions?
Catalog NumberQuantity
44323341 x 96-well Plate
Catalog number 4432334
Price (USD)
177.00
Each
-
Add to cart
Quantity:
1 x 96-well Plate
Price (USD)
177.00
Each
Add to cart
Ask our AI about this Product
This VIC™ dye spectral calibration plate can be used to maintain your ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, or QuantStudio™ 12K Flex system with 96-well block. Calibration and verification should be run at least every six months. Please see your instrument maintenance guide for full instructions on use.

The dye formulations for your real-time PCR system will improve results with multiplexing by more accurately representing fluorescent spectra found in your real-time PCR experiments.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Calibration TypeSpectral Calibration
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)QuantStudio™ 12 K Flex, QuantStudio™ 6 Flex, QuantStudio™ 7, ViiA™ 7 System
Quantity1 x 96-well Plate
Shipping ConditionDry Ice
Product TypeDye Spectral Calibration Plate
Unit SizeEach
Contents & Storage
Store at -15°C to -25°C
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2505211Certificate of AnalysisJun 19, 20254432334
2504210Certificate of AnalysisApr 22, 20254432334
2502209Certificate of AnalysisMar 10, 20254432334
2501208Certificate of AnalysisFeb 12, 20254432334
2401192Certificate of AnalysisJan 27, 20254432334
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

To open the software download on your Mac, you will need to adjust your security settings using the following instructions from Apple support:

Safely open apps on your mac

Please note, the software should run on Mac OS 10.10 or later.

Find additional tips, troubleshooting help, and resources within our QuantStudio 3 and QuantStudio 5 Real Time PCR Systems Support Center.

You can get a summary of the calibrations from the QuantStudio Software by going to Instrument > Instrument Console > Manage Instrument > Information > Print Calibration Log.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

No, you do not need to back up the calibration files as all of the calibration files are stored on the ViiA7 Real-Time PCR instrument. Once the computer software is properly installed and the computer is connected to the instrument, the calibration information will automatically update in the QuantStudio Software.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Yes, you can still start a run on the ViiA7 Real-Time PCR System if the expired calibration notice displays. This is just a reminder and you can close it to proceed with the run. Additionally, you can change when you want the notifications to show up and how frequently you want to be notified to calibrate the instrument. To do this, go to the toolbar in the QuantStudio Software and select Instrument > Instrument Console. Select your instrument icon and click on Manager Instrument > Maintenance > Calibration Reminders. From the Calibration reminders Settings page you can update when the calibrations expire, turn expiration reminders on/off, and set when the reminders show up.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Share catalog number, name or link

1x1 image pixel for data collection