Thermo Scientific Chemicals

2-Aminophenylboronic acid hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 445310010
1 g, Each
Thermo Scientific Chemicals

2-Aminophenylboronic acid hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 445310010
1 g, Each
Quantity
Catalog number: 445310010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
863753-30-4
IUPAC Name
(2-aminophenyl)boronic acid hydrochloride
Molecular Formula
C6H9BClNO2
InChI Key
WPDASZCYRKGSTO-UHFFFAOYSA-N
SMILES
Cl.NC1=CC=CC=C1B(O)O
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
HPLC
>=96.0 %
NMR (1H-NMR)
Consistent with structure
Appearance (Form)
Powder or solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text