Sodium bromide, 99+%, for biochemistry
Sodium bromide, 99+%, for biochemistry
Sodium bromide, 99+%, for biochemistry
Sodium bromide, 99+%, for biochemistry
Thermo Scientific Chemicals

Sodium bromide, 99+%, for biochemistry

CAS: 7647-15-6 | BrNa | 102.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 446805000
Price (USD)
113.65
Special offer
Online exclusive
134.00
Save 20.35 (15%)
Each
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
113.65
Special offer
Online exclusive
134.00
Save 20.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7647-15-6
IUPAC Namesodium bromide
Molecular FormulaBrNa
InChI KeyJHJLBTNAGRQEKS-UHFFFAOYSA-M
SMILES[Na+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Magnesium (Mg)=<0.001 %
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration Argentometric>=99.0 %
pH5.0 to 8.8
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0454372Certificate of AnalysisJan 31, 2025446801000, 446805000
A0464746Certificate of AnalysisAug 01, 2024446801000, 446805000
A0461937Certificate of AnalysisApr 12, 2024446801000, 446805000
A0454372Certificate of AnalysisAug 25, 2023446801000, 446805000
A0449515Certificate of AnalysisFeb 27, 2023446801000, 446805000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection