1,2-Propanediol, ACS reagent
1,2-Propanediol, ACS reagent
1,2-Propanediol, ACS reagent
1,2-Propanediol, ACS reagent
Thermo Scientific Chemicals

1,2-Propanediol, ACS reagent

CAS: 57-55-6 | C3H8O2 | 76.09 g/mol
数量:
100 mL
500 mL
2.5 L
製品番号(カタログ番号) 447415000
価格(JPY)
-
見積もりを依頼する
数量:
500 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS57-55-6
IUPAC Namepropane-1,2-diol
Molecular FormulaC3H8O2
InChI KeyDNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILESCC(O)CO
さらに表示
Water=<0.2 %
Chloride (Cl)=<1 ppm
Residue after ignition=<0.005 %
Color scale=<10 APHA
Appearance (Form)Clear viscous liquid
さらに表示
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

ドキュメントおよびダウンロード

証明書

ロット番号Certificate TypeDateCatalog Number(s)
A0473680Certificate of Analysis2025年6月30日447410025, 447411000, 447415000
A0470983Certificate of Analysis2025年5月17日447410025, 447411000, 447415000
A0467774Certificate of Analysis2025年1月31日447410025, 447411000, 447415000
A0456215Certificate of Analysis2023年9月21日447410025, 447411000, 447415000
A0453078Certificate of Analysis2023年5月11日447410025, 447411000, 447415000
5件の結果が表示されました。 上記から特定の証明書を検索します

Safety Data Sheets

こちらの製品には複数の製品番号が設定されています。

1x1 image pixel for data collection