Sodium phosphate, dibasic dihydrate, 98%, extra pure
Sodium phosphate, dibasic dihydrate, 98%, extra pure
Thermo Scientific Chemicals

Sodium phosphate, dibasic dihydrate, 98%, extra pure

CAS: 10028-24-7 | H5Na2O6P | 177.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Catalog number 448150050
Price (USD)
164.00
Each
In stock
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (USD)
164.00
Each
Add to cart
Chemical Identifiers
CAS10028-24-7
IUPAC Namedisodium dihydrate hydrogen phosphate
Molecular FormulaH5Na2O6P
InChI KeyKDQPSPMLNJTZAL-UHFFFAOYSA-L
SMILESO.O.[Na+].[Na+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white or colorless
Appearance (Form)Crystalline powder or crystals
Titration with HClO497.5 to 102.5 % (dried substance)
Solubility(5 % in water) Clear colorless
Heavy metals (as Pb)=<20 ppm
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0469291Certificate of AnalysisJan 30, 2025448150010, 448150050
A0466260Certificate of AnalysisSep 25, 2024448150010, 448150050
A0462340Certificate of AnalysisMay 01, 2024448150010, 448150050
A0459283Certificate of AnalysisJan 15, 2024448150010, 448150050
A0457111Certificate of AnalysisNov 03, 2023448150010, 448150050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection