Itraconazole, 99%
Itraconazole, 99%
Itraconazole, 99%
Thermo Scientific Chemicals

Itraconazole, 99%

Itraconazole, or voriconazole, CAS # 84625-61-6, is an N-arylpiperazine that has antifungal properties. | CAS: 84625-61-6 | C35H38Cl2N8O4 | 705.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 452870050
Price (USD)
125.65
Online Exclusive
139.00
Save 13.35 (10%)
Each
-
Add to cart
Quantity:
5 g
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
125.65
Online Exclusive
139.00
Save 13.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS84625-61-6
IUPAC Name1-(butan-2-yl)-4-{4-[4-(4-{[(2R,4S)-2-(2,4-dichlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy}phenyl)piperazin-1-yl]phenyl}-4,5-dihydro-1H-1,2,4-triazol-5-one
Molecular FormulaC35H38Cl2N8O4
InChI KeyVHVPQPYKVGDNFY-ZPGVKDDISA-N
SMILESCCC(C)N1N=CN(C1=O)C1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(OC[C@H]2CO[C@@](CN3C=NC=N3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point165°C to 169°C
Appearance (Color)White to almost white
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=98.5 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Itraconazole is an aromatic ether that works as a strong inhibitor of P-glycoproteins and CYP3A4

• Itraconazole also acts as a hedgehog signaling inhibitor and P450 inhibitor

Applications

• Itraconazole can be exhibits antifungal activity against organisms including aspergillosis, blastomycosis, candidiasis, chromoblastomycosis, coccidioidomycosis, cryptococcosis, histoplasmosis, and sporotrichosis

• Itraconazole may be used to experimentally target the delivery of other molecules, such as plasmids, to cells

RUO – Research Use Only

General References:

  1. Sharma, P.; Bhalla, M.; Thami, G.P.; Chander, J. Evaluation of efficacy and safety of oral terbinafine and itraconazole combination therapy in the management of dermatophytosis. J Dermatolog Treat. 2020, 31(7), 749-753.
  2. Hamed Khater, M.; Khattab, F.M. Combined long-pulsed Nd-Yag laser and itraconazole versus itraconazole alone in the treatment of onychomycosis nails. J Dermatolog Treat. 2020, 31(4), 406-409.
Have questions about this product? Ask our AI assisted search.
I am getting lower yield than I expected with the ExpiCHO expression system. What should I do to improve my results?
I don't have ExpiCHO-S cells. Can Expifectamine CHO transfection reagent and enhancer be used with other CHO cell lines than the ExpiCHO-S cell line?
What type of Rabbit IgG does the Antibody-Expressing Positive Control Vector (Cat. No. A14662) express?
Do I need to change the media after the ExpiFectamine CHO transfection?
How does the protein expression yield of the ExpiCHO expression system compare to FreeStyle CHO or Expi293 expression systems?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0450186Certificate of AnalysisMay 02, 2023452870050, 452870250
A0400563Certificate of AnalysisDec 14, 2022452870050, 452870250
A0398764Certificate of AnalysisDec 14, 2022452870050, 452870250
A0399018Certificate of AnalysisDec 14, 2022452870050, 452870250
A0448248Certificate of AnalysisDec 13, 2022452870050, 452870250
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection