Lead(II) sulfide, 90%, pure
Lead(II) sulfide, 90%, pure
Lead(II) sulfide, 90%, pure
Lead(II) sulfide, 90%, pure
Thermo Scientific Chemicals

Lead(II) sulfide, 90%, pure

CAS: 1314-87-0 | PbS | 239.30 g/mol
製品番号(カタログ番号) 453970010
価格(JPY)
-
見積もりを依頼する
数量:
1 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1314-87-0
IUPAC Nameλ²-lead(2+) sulfanediide
Molecular FormulaPbS
InChI KeyFILXVLQXZYXBSJ-UHFFFAOYSA-N
SMILES[S--].[Pb++]
さらに表示
Appearance (Color)Grey to purple
Assay>=78 % (Pb)
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

こちらの製品には複数の製品番号が設定されています。