2,3-Diaminobenzoic acid, 95%
2,3-Diaminobenzoic acid, 95%
2,3-Diaminobenzoic acid, 95%
2,3-Diaminobenzoic acid, 95%
Thermo Scientific Chemicals

2,3-Diaminobenzoic acid, 95%

CAS: 603-81-6 | C7H8N2O2 | 152.15 g/mol
製品番号(カタログ番号) 457490050
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS603-81-6
Appearance (Color)Off-white to dark brown to black
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=94.0 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

ドキュメントおよびダウンロード

証明書

ロット番号Certificate TypeDateCatalog Number(s)
341952Certificate of Analysis2022年5月14日457490010, 457490050
1件の結果が表示されました。 上記から特定の証明書を検索します

Safety Data Sheets

こちらの製品には複数の製品番号が設定されています。

1x1 image pixel for data collection