Pentamidine isethionate salt, 97%
Pentamidine isethionate salt, 97%
Pentamidine isethionate salt, 97%
Pentamidine isethionate salt, 97%
Thermo Scientific Chemicals

Pentamidine isethionate salt, 97%

CAS: 140-64-7 | C19H24N4O2·2C2H6O4S | 592.68 g/mol
Have Questions?
Catalog NumberQuantity
4618600101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 461860010
Price (EUR)
134,30
Online Exclusive
158,00
Save 23,70 (15%)
Each
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
134,30
Online Exclusive
158,00
Save 23,70 (15%)
Each
Add to cart
Chemical Identifiers
CAS140-64-7
IUPAC Namebis(2-hydroxyethane-1-sulfonic acid); 4-{[5-(4-carbamimidoylphenoxy)pentyl]oxy}benzene-1-carboximidamide
Molecular FormulaC23H36N4O10S2
InChI KeyYBVNFKZSMZGRAD-UHFFFAOYSA-N
SMILESOCCS(O)(=O)=O.OCCS(O)(=O)=O.NC(=N)C1=CC=C(OCCCCCOC2=CC=C(C=C2)C(N)=N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point188°C to 194°C
HPLC>=96.0 %
Infrared spectrumConforms
Appearance (Color)White to off-white
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470228Certificate of AnalysisMay 17, 2025461860010
A0464417Certificate of AnalysisJul 26, 2024461860010
A0459505Certificate of AnalysisFeb 01, 2024461860010
N16J042Certificate of AnalysisMay 08, 2023461860010
A0442871Certificate of AnalysisJun 14, 2022461860010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Skin Sensitization
Category 1
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Warning

Hazard Statements

H315 - Causes skin irritation

H317 - May cause an allergic skin reaction

H319 - Causes serious eye irritation

H335 - May cause respiratory irritation

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P312 - Call a POISON CENTER or doctor if you feel unwell

P332 + P313 - If skin irritation occurs: Get medical advice/attention

P337 + P313 - If eye irritation persists: Get medical advice/attention

Share catalog number, name or link

1x1 image pixel for data collection