Citalopram hydrobromide
Citalopram hydrobromide
Citalopram hydrobromide
Citalopram hydrobromide
Thermo Scientific Chemicals

Citalopram hydrobromide

CAS: 59729-32-7 | C20H21FN2O·HBr | 405.3 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 462310050
Price (USD)
184.65
Special offer
Online exclusive
217.00
Save 32.35 (15%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
184.65
Special offer
Online exclusive
217.00
Save 32.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS59729-32-7
IUPAC Namehydrogen 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-2-benzofuran-5-carbonitrile bromide
Molecular FormulaC20H22BrFN2O
InChI KeyWIHMBLDNRMIGDW-UHFFFAOYNA-N
SMILES[H+].[Br-].CN(C)CCCC1(OCC2=CC(=CC=C12)C#N)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<0.5 % (1 g, 105°C, 4 h)
HPLC>=97.5 %
Solubility(1 % in ethanol) Clear colorless
Appearance (Color)White to almost white
Appearance (Form)Powder
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0469931Certificate of AnalysisMay 17, 2025462310010, 462310050
A0471751Certificate of AnalysisMay 17, 2025462310010, 462310050
A0467920Certificate of AnalysisJan 25, 2025462310010, 462310050
A0464196Certificate of AnalysisJul 11, 2024462310010, 462310050
A0456320Certificate of AnalysisOct 04, 2023462310010, 462310050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection