Pazopanib
Thermo Scientific Chemicals

Pazopanib

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 462380050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS444731-52-6
IUPAC Name5-({4-[(2,3-dimethyl-2H-indazol-6-yl)(methyl)amino]pyrimidin-2-yl}amino)-2-methylbenzene-1-sulfonamide
Molecular FormulaC21H23N7O2S
InChI KeyCUIHSIWYWATEQL-UHFFFAOYSA-N
SMILESCN(C1=CC2=NN(C)C(C)=C2C=C1)C1=NC(NC2=CC(=C(C)C=C2)S(N)(=O)=O)=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light yellow or beige
Appearance (Form)Powder
Infrared spectrumConforms
Solubility(0.5 % in DMSO, warmed) Clear solution
HPLC>=97.5 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0386094Certificate of AnalysisMay 17, 2025462380050, 462380010
A0386094Certificate of AnalysisSep 05, 2023462380050, 462380010
2 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection