Esmolol hydrochloride
Product Image
Thermo Scientific Chemicals

Esmolol hydrochloride

製品番号(カタログ番号) 462460250
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS81161-17-3
Appearance (Color)White to off-white
Appearance (Form)Powder
Melting point86°C to 92°C
HPLC>=97.5 %
Infrared spectrumConforms
さらに表示
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

ドキュメントおよびダウンロード

証明書

ロット番号Certificate TypeDateCatalog Number(s)
A0386110Certificate of Analysis2025年5月17日462460050, 462460250
A0386110Certificate of Analysis2022年9月19日462460050, 462460250
2件の結果が表示されました。 上記から特定の証明書を検索します

Safety Data Sheets

こちらの製品には複数の製品番号が設定されています。

1x1 image pixel for data collection