Dexmedetomidine hydrochloride
Product Image
Thermo Scientific Chemicals

Dexmedetomidine hydrochloride

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 465432500
Price (USD)
163.65
Online Exclusive
182.00
Save 18.35 (10%)
Each
In stock
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
163.65
Online Exclusive
182.00
Save 18.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS145108-58-3
IUPAC Namehydrogen 5-[(1S)-1-(2,3-dimethylphenyl)ethyl]-1H-imidazole chloride
Molecular FormulaC13H17ClN2
InChI KeyVPNGEIHDPSLNMU-MERQFXBCSA-N
SMILES[H+].[Cl-].C[C@H](C1=CN=CN1)C1=CC=CC(C)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Powder
HPLC>=97.5 %
Specific optical rotation+48° to +54° (20°C, 589 nm) (1% in water)
Infrared spectrumConforms
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471996Certificate of AnalysisMay 17, 2025465430010, 465432500
A0463571Certificate of AnalysisJun 12, 2024465430010, 465432500
A0410525Certificate of AnalysisApr 11, 2022465430010, 465432500
3 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection