Rutaecarpine
Thermo Scientific Chemicals

Rutaecarpine

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 465982500
Price (USD)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS84-26-4
IUPAC Name3,13,21-triazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(10),4,6,8,15,17,19-octaen-14-one
Molecular FormulaC18H13N3O
InChI KeyACVGWSKVRYFWRP-UHFFFAOYSA-N
SMILESO=C1N2CCC3=C(NC4=CC=CC=C34)C2=NC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Color)White to beige
Appearance (Form)Powder
Loss on drying=<0.5 %
Infrared spectrumConforms
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0416787Certificate of AnalysisMay 17, 2025465980010, 465982500
A0416787Certificate of AnalysisOct 10, 2022465980010, 465982500
2 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection