Thermo Scientific™
Finnpipette™ F1 Multichannel Pipettes
Catalog number: 4661040N
Have Questions?
Thermo Scientific™
Finnpipette™ F1 Multichannel Pipettes
Catalog number: 4661040N
Have Questions?
Get optimal performance and ergonomics from the Finnpipette™ F1 system, which delivers performance with precision, all in a lightweight design that maximizes user comfort and reduces strain. It’s the ideal system for applications using expensive reagents and valuable samples.
Catalog Number
4661040N
Unit Size
Each
Number of Channels
12
Volume (Metric)
1 to 10 μL
Compatible Tips
Finntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
Color
Pink
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
DescriptionFinnpipette F1
ColorPink
Number of Channels12
Volume (Metric)1 to 10 μL
Increments0.02μL
Color CodePink
Unit SizeEach
Catalog Number
Unit Size
Each
Number of Channels
8
Volume (Metric)
1 to 10 μL
Compatible Tips
Finntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
Color
Pink
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
DescriptionFinnpipette F1
ColorPink
Number of Channels8
Volume (Metric)1 to 10 μL
Increments0.02μL
Color CodePink
Unit SizeEach
Catalog Number
Unit Size
Each
Number of Channels
8
Volume (Metric)
5 to 50 μL
Compatible Tips
Finntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext
Color
Yellow
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext
DescriptionFinnpipette F1
ColorYellow
Number of Channels8
Volume (Metric)5 to 50 μL
Increments0.1μL
Color CodeYellow
Unit SizeEach
Catalog Number
Unit Size
Each
Number of Channels
8
Volume (Metric)
10 to 100 μL
Compatible Tips
Finntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide
Color
Yellow
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide
DescriptionFinnpipette F1
ColorYellow
Number of Channels8
Volume (Metric)10 to 100 μL
Increments0.2μL
Color CodeYellow
Unit SizeEach
Catalog Number
Unit Size
Each
Number of Channels
8
Volume (Metric)
30 to 300 μL
Compatible Tips
Finntip 300, Filter 300, Flex 300, Flex Filter 300
Color
Orange
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 300, Filter 300, Flex 300, Flex Filter 300
DescriptionFinnpipette F1
ColorOrange
Number of Channels8
Volume (Metric)30 to 300 μL
Increments1μL
Color CodeOrange
Unit SizeEach
Catalog Number
Unit Size
Each
Number of Channels
12
Volume (Metric)
5 to 50 μL
Compatible Tips
Finntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext
Color
Yellow
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext
DescriptionFinnpipette F1
ColorYellow
Number of Channels12
Volume (Metric)5 to 50 μL
Increments0.1μL
Color CodeYellow
Unit SizeEach
Catalog Number
Unit Size
Each
Number of Channels
12
Volume (Metric)
10 to 100 μL
Compatible Tips
Finntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide
Color
Yellow
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide
DescriptionFinnpipette F1
ColorYellow
Number of Channels12
Volume (Metric)10 to 100 μL
Increments0.2μL
Color CodeYellow
Unit SizeEach
Catalog Number
Unit Size
Each
Number of Channels
12
Volume (Metric)
30 to 300 μL
Compatible Tips
Finntip 300, Filter 300, Flex 300, Flex Filter 300
Color
Orange
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 300, Filter 300, Flex 300, Flex Filter 300
DescriptionFinnpipette F1
ColorOrange
Number of Channels12
Volume (Metric)30 to 300 μL
Increments1μL
Color CodeOrange
Unit SizeEach
Catalog Number
Unit Size
Each
Number of Channels
16
Volume (Metric)
1 to 10 μL
Compatible Tips
Finntip 20 Micro, Filter 20 Micro
Color
Purple
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 20 Micro, Filter 20 Micro
DescriptionFinnpipette F1
ColorPurple
Number of Channels16
Volume (Metric)1 to 10 μL
Increments0.02μL
Color CodePurple
Unit SizeEach
Catalog Number
Unit Size
Each
Number of Channels
16
Volume (Metric)
5 to 50 μL
Compatible Tips
Finntip 50 Micro, Filter 50 Micro
Color
Turquoise Button
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 50 Micro, Filter 50 Micro
DescriptionFinnpipette F1
ColorTurquoise Button
Number of Channels16
Volume (Metric)5 to 50 μL
Increments0.1μL
Color CodeTurquoise
Unit SizeEach
Showing 10 of 10
Catalog NumberSpecificationsUnit SizeNumber of ChannelsVolume (Metric)Compatible TipsColorPrice (USD)AvailabilityQuantity
4661040NFull specifications
Each121 to 10 μLFinntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10Pink
-
Compatible TipsFinntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
DescriptionFinnpipette F1
ColorPink
Number of Channels12
Volume (Metric)1 to 10 μL
Increments0.02μL
Color CodePink
Unit SizeEach
4661000NFull specifications
Each81 to 10 μLFinntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10Pink
-
Compatible TipsFinntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
DescriptionFinnpipette F1
ColorPink
Number of Channels8
Volume (Metric)1 to 10 μL
Increments0.02μL
Color CodePink
Unit SizeEach
4661010NFull specifications
Each85 to 50 μLFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 ExtYellow
-
Compatible TipsFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext
DescriptionFinnpipette F1
ColorYellow
Number of Channels8
Volume (Metric)5 to 50 μL
Increments0.1μL
Color CodeYellow
Unit SizeEach
4661020NFull specifications
Each810 to 100 μLFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 WideYellow
-
Compatible TipsFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide
DescriptionFinnpipette F1
ColorYellow
Number of Channels8
Volume (Metric)10 to 100 μL
Increments0.2μL
Color CodeYellow
Unit SizeEach
4661030NFull specifications
Each830 to 300 μLFinntip 300, Filter 300, Flex 300, Flex Filter 300Orange
-
Compatible TipsFinntip 300, Filter 300, Flex 300, Flex Filter 300
DescriptionFinnpipette F1
ColorOrange
Number of Channels8
Volume (Metric)30 to 300 μL
Increments1μL
Color CodeOrange
Unit SizeEach
4661050NFull specifications
Each125 to 50 μLFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 ExtYellow
-
Compatible TipsFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext
DescriptionFinnpipette F1
ColorYellow
Number of Channels12
Volume (Metric)5 to 50 μL
Increments0.1μL
Color CodeYellow
Unit SizeEach
4661060NFull specifications
Each1210 to 100 μLFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 WideYellow
-
Compatible TipsFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide
DescriptionFinnpipette F1
ColorYellow
Number of Channels12
Volume (Metric)10 to 100 μL
Increments0.2μL
Color CodeYellow
Unit SizeEach
4661070NFull specifications
Each1230 to 300 μLFinntip 300, Filter 300, Flex 300, Flex Filter 300Orange
-
Compatible TipsFinntip 300, Filter 300, Flex 300, Flex Filter 300
DescriptionFinnpipette F1
ColorOrange
Number of Channels12
Volume (Metric)30 to 300 μL
Increments1μL
Color CodeOrange
Unit SizeEach
4661080NFull specifications
Each161 to 10 μLFinntip 20 Micro, Filter 20 MicroPurple
-
Compatible TipsFinntip 20 Micro, Filter 20 Micro
DescriptionFinnpipette F1
ColorPurple
Number of Channels16
Volume (Metric)1 to 10 μL
Increments0.02μL
Color CodePurple
Unit SizeEach
4661090NFull specifications
Each165 to 50 μLFinntip 50 Micro, Filter 50 MicroTurquoise Button
-
Compatible TipsFinntip 50 Micro, Filter 50 Micro
DescriptionFinnpipette F1
ColorTurquoise Button
Number of Channels16
Volume (Metric)5 to 50 μL
Increments0.1μL
Color CodeTurquoise
Unit SizeEach
Showing 10 of 10
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F1: Everything you want in your everyday, go-to pipette
Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI
Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μl models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
5 Year Warranty
With convenient online registration, and may vary by country.
Learn more about the complete Finnpipette family >
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov