Iohexol
Thermo Scientific Chemicals

Iohexol

CAS: 66108-95-0 | C19H26I3N3O9, C19H26I3N3O9 | 821.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 466651000
Price (USD)
382.65
Online Exclusive
425.00
Save 42.35 (10%)
Each
In stock
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
382.65
Online Exclusive
425.00
Save 42.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS66108-95-0
IUPAC NameN1,N3-bis(2,3-dihydroxypropyl)-5-[N-(2,3-dihydroxypropyl)acetamido]-2,4,6-triiodobenzene-1,3-dicarboxamide
Molecular FormulaC19H26I3N3O9
InChI KeyNTHXOOBQLCIOLC-UHFFFAOYNA-N
SMILESCC(=O)N(CC(O)CO)C1=C(I)C(C(=O)NCC(O)CO)=C(I)C(C(=O)NCC(O)CO)=C1I
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=98.0 % (sum of isomers)
Water=<4.0 % (K.F.)
Appearance (Color)White to almost white
Appearance (Form)Powder
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471259Certificate of AnalysisMay 17, 2025466651000, 466650250
A0468668Certificate of AnalysisJan 25, 2025466651000, 466650250
A0465050Certificate of AnalysisAug 14, 2024466651000, 466650250
A0463357Certificate of AnalysisJun 14, 2024466651000, 466650250
A0460900Certificate of AnalysisMar 14, 2024466651000, 466650250
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection