Thermo Scientific™
Heratherm™ Advanced Protocol Ovens, 230VAC 60Hz
Catalog number: 51028144
Have Questions?
Thermo Scientific™
Heratherm™ Advanced Protocol Ovens, 230VAC 60Hz
Catalog number: 51028144
Have Questions?
Carry out multiple applications with Thermo Scientific™ Heratherm™ Advanced Protocol Ovens, which include a sophisticated timer, precise temperature uniformity and low energy-consumption. Featuring either mechanical or gravity convection technology with excellent temperature values, This flexible tool for the lab may be used for multiple applications.
Catalog Number
51028144
Unit Size
Each
Capacity (Metric)
168 L
Type
Gravity Convection
Material
Coated Steel
Plug Type
Country-specific
Price (USD)
Full specifications
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Description51028144
Capacity (Metric)168 L
Temperature (Metric)330°C
TypeGravity Convection
MaterialCoated Steel
No. of Shelves2 supplied/19 max.
Plug TypeCountry-specific
Temperature Stability±2.5°C
Dimensions (D x W x H) Interior22.2 x 17.2 x 26.8 in. (56.4 x 43.8 x 68 cm)
Dimensions (L x W x H)29.1 x 25.2 x 36.2 in. (73.8 x 64 x 92 cm)
Length (English)29.1 in.
Length (Metric)73.8 cm
Width (English)25.2 in.
Width (English) Interior17.2 in.
Width (Metric)64 cm
Width (Metric) Interior43.8 cm
Height (English)36.2 in.
Height (English) Interior26.8 in.
Height (Metric)92 cm
Height (Metric) Interior68 cm
Depth (English) Interior22.2 in.
Depth (Metric) Interior56.4 cm
Electrical Requirements230 V 50/60 Hz
Voltage230 V
Wattage3100 w
Amperage13.5 A
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Capacity (Metric)
61 L
Type
Gravity Convection
Material
Coated Steel
Plug Type
Country-specific
Price (USD)
Full specifications
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Description51028142
Capacity (Metric)61 L
Temperature (Metric)330°C
TypeGravity Convection
MaterialCoated Steel
No. of Shelves2 supplied/13 max.
Plug TypeCountry-specific
Temperature Stability±2.5°C
Dimensions (D x W x H) Interior15.3 x 12.9 x 18.9 in. (38.9 x 32.8 x 48 cm)
Dimensions (L x W x H)22.2 x 20.9 x 28.3 in. (56.5 x 53 x 72 cm)
Length (English)22.2 in.
Length (Metric)56.5 cm
Width (English)20.9 in.
Width (English) Interior12.9 in.
Width (Metric)53 cm
Width (Metric) Interior32.8 cm
Height (English)28.3 in.
Height (English) Interior18.9 in.
Height (Metric)72 cm
Height (Metric) Interior48 cm
Depth (English) Interior15.3 in.
Depth (Metric) Interior38.9 cm
Electrical Requirements230 V 50/60 Hz
Voltage230 V
Wattage1800 w
Amperage7.9 A
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Capacity (Metric)
62 L
Type
Mechanical Convection Oven
Material
Coated Steel
Plug Type
Country-specific
Price (USD)
Full specifications
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Description51028151
Capacity (Metric)62 L
Temperature (Metric)330°C
TypeMechanical Convection Oven
MaterialCoated Steel
No. of Shelves2 supplied/13 max.
Plug TypeCountry-specific
Temperature Stability±1.8°C
Dimensions (D x W x H) Interior13.5 x 13.9 x 20 in. (34.3 x 35.4 x 50.8 cm)
Dimensions (L x W x H)22.2 x 20.9 x 28.3 in. (56.5 x 53 x 72 cm)
Length (English)22.2 in.
Length (Metric)56.5 cm
Width (English)20.9 in.
Width (English) Interior13.9 in.
Width (Metric)53 cm
Width (Metric) Interior35.4 cm
Height (English)28.3 in.
Height (English) Interior20 in.
Height (Metric)72 cm
Height (Metric) Interior50.8 cm
Depth (English) Interior13.5 in.
Depth (Metric) Interior34.3 cm
Electrical Requirements230 V 50/60 Hz
Voltage230 V
Wattage1400 w
Amperage6.1 A
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Capacity (Metric)
97 L
Type
Mechanical Convection Oven
Material
Coated Steel
Plug Type
Country-specific
Price (USD)
Full specifications
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Description51028152
Capacity (Metric)97 L
Temperature (Metric)330°C
TypeMechanical Convection Oven
MaterialCoated Steel
No. of Shelves2 supplied/16 max.
Plug TypeCountry-specific
Temperature Stability±1.3°C
Dimensions (D x W x H) Interior13.5 x 18.3 x 23.9 in. (34.3 x 46.4 x 60.8 cm)
Dimensions (L x W x H)22.2 x 25.2 x 32.3 in. (56.5 x 64 x 82 cm)
Length (English)22.2 in.
Length (Metric)56.5 cm
Width (English)25.2 in.
Width (English) Interior18.3 in.
Width (Metric)64 cm
Width (Metric) Interior46.4 cm
Height (English)32.3 in.
Height (English) Interior23.9 in.
Height (Metric)82 cm
Height (Metric) Interior60.8 cm
Depth (English) Interior13.5 in.
Depth (Metric) Interior34.3 cm
Electrical Requirements230 V 50/60 Hz
Voltage230 V
Wattage3060 w
Amperage13.3 A
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Capacity (Metric)
170 L
Type
Mechanical Convection Oven
Material
Coated Steel
Plug Type
Country-specific
Price (USD)
Full specifications
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Description51028153
Capacity (Metric)170 L
Temperature (Metric)330°C
TypeMechanical Convection Oven
MaterialCoated Steel
No. of Shelves2 supplied/19 max.
Plug TypeCountry-specific
Temperature Stability±1.8°C
Dimensions (D x W x H) Interior20.4 x 18.3 x 27.9 in. (51.8 x 46.4 x 70.8 cm)
Dimensions (L x W x H)29.1 x 25.2 x 36.2 in. (73.8 x 64 x 92 cm)
Length (English)29.1 in.
Length (Metric)73.8 cm
Width (English)25.2 in.
Width (English) Interior18.3 in.
Width (Metric)64 cm
Width (Metric) Interior46.4 cm
Height (English)36.2 in.
Height (English) Interior27.9 in.
Height (Metric)92 cm
Height (Metric) Interior70.8 cm
Depth (English) Interior20.4 in.
Depth (Metric) Interior51.8 cm
Electrical Requirements230 V 50/60 Hz
Voltage230 V
Wattage3060 w
Amperage13.3 A
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Capacity (Metric)
61 L
Type
Gravity Convection
Material
Stainless Steel
Plug Type
Country-specific
Price (USD)
Full specifications
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Description51028709
Capacity (Metric)61 L
Temperature (Metric)330°C
TypeGravity Convection
MaterialStainless Steel
No. of Shelves2 supplied/13 max.
Plug TypeCountry-specific
Temperature Stability±2.5°C
Dimensions (D x W x H) Interior15.3 x 12.9 x 18.9 in. (38.9 x 32.8 x 48 cm)
Dimensions (L x W x H)22.2 x 20.9 x 28.3 in. (56.5 x 53 x 72 cm)
Length (English)22.2 in.
Length (Metric)56.5 cm
Width (English)20.9 in.
Width (English) Interior12.9 in.
Width (Metric)53 cm
Width (Metric) Interior32.8 cm
Height (English)28.3 in.
Height (English) Interior18.9 in.
Height (Metric)72 cm
Height (Metric) Interior48 cm
Depth (English) Interior15.3 in.
Depth (Metric) Interior38.9 cm
Electrical Requirements230 V 50/60 Hz
Voltage230 V
Wattage1800 w
Amperage7.9 A
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Capacity (Metric)
62 L
Type
Mechanical Convection Oven
Material
Stainless Steel
Plug Type
Country-specific
Price (USD)
Full specifications
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Description51028713
Capacity (Metric)62 L
Temperature (Metric)330°C
TypeMechanical Convection Oven
MaterialStainless Steel
No. of Shelves2 supplied/13 max.
Plug TypeCountry-specific
Temperature Stability±1.8°C
Dimensions (D x W x H) Interior13.5 x 13.9 x 20 in. (34.3 x 35.4 x 50.8 cm)
Dimensions (L x W x H)22.2 x 20.9 x 28.3 in. (56.5 x 53 x 72 cm)
Length (English)22.2 in.
Length (Metric)56.5 cm
Width (English)20.9 in.
Width (English) Interior13.9 in.
Width (Metric)53 cm
Width (Metric) Interior35.4 cm
Height (English)28.3 in.
Height (English) Interior20 in.
Height (Metric)72 cm
Height (Metric) Interior50.8 cm
Depth (English) Interior13.5 in.
Depth (Metric) Interior34.3 cm
Electrical Requirements230 V 50/60 Hz
Voltage230 V
Wattage1400 w
Amperage6.1 A
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Capacity (Metric)
97 L
Type
Mechanical Convection Oven
Material
Stainless Steel
Plug Type
Country-specific
Price (USD)
Full specifications
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Description51028715
Capacity (Metric)97 L
Temperature (Metric)330°C
TypeMechanical Convection Oven
MaterialStainless Steel
No. of Shelves2 supplied/16 max.
Plug TypeCountry-specific
Temperature Stability±1.3°C
Dimensions (D x W x H) Interior13.5 x 18.3 x 23.9 in. (34.3 x 46.4 x 60.8 cm)
Dimensions (L x W x H)22.2 x 25.2 x 32.3 in. (56.5 x 64 x 82 cm)
Length (English)22.2 in.
Length (Metric)56.5 cm
Width (English)25.2 in.
Width (English) Interior18.3 in.
Width (Metric)64 cm
Width (Metric) Interior46.4 cm
Height (English)32.3 in.
Height (English) Interior23.9 in.
Height (Metric)82 cm
Height (Metric) Interior60.8 cm
Depth (English) Interior13.5 in.
Depth (Metric) Interior34.3 cm
Electrical Requirements230 V 50/60 Hz
Voltage230 V
Wattage3060 w
Amperage13.3 A
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Capacity (Metric)
170 L
Type
Mechanical Convection Oven
Material
Stainless Steel
Plug Type
Country-specific
Price (USD)
Full specifications
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Description51028716
Capacity (Metric)170 L
Temperature (Metric)330°C
TypeMechanical Convection Oven
MaterialStainless Steel
No. of Shelves2 supplied/19 max.
Plug TypeCountry-specific
Temperature Stability±1.8°C
Dimensions (D x W x H) Interior20.4 x 18.3 x 27.9 in. (51.8 x 46.4 x 70.8 cm)
Dimensions (L x W x H)29.1 x 25.2 x 36.2 in. (73.8 x 64 x 92 cm)
Length (English)29.1 in.
Length (Metric)73.8 cm
Width (English)25.2 in.
Width (English) Interior18.3 in.
Width (Metric)64 cm
Width (Metric) Interior46.4 cm
Height (English)36.2 in.
Height (English) Interior27.9 in.
Height (Metric)92 cm
Height (Metric) Interior70.8 cm
Depth (English) Interior20.4 in.
Depth (Metric) Interior51.8 cm
Electrical Requirements230 V 50/60 Hz
Voltage230 V
Wattage3060 w
Amperage13.3 A
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Capacity (Metric)
396 L
Type
Mechanical Convection Oven
Material
Stainless Steel
Plug Type
Country-specific
Price (USD)
Full specifications
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Description51029332
Capacity (Metric)396 L
Temperature (Metric)330°C
TypeMechanical Convection Oven
MaterialStainless Steel
No. of Shelves2 supplied/39 max.
Plug TypeCountry-specific
Temperature Stability±2.1°C
Dimensions (D x W x H) Interior21.5 x 21.4 x 52.6 in. (54.5 x 54.4 x 133.5 cm)
Dimensions (L x W x H)30.3 x 30.6 x 60.8 in. (77 x 77.8 x 154.5 cm)
Length (English)30.3 in.
Length (Metric)77 cm
Width (English)30.6 in.
Width (English) Interior21.4 in.
Width (Metric)77.8 cm
Width (Metric) Interior54.4 cm
Height (English)60.8 in.
Height (English) Interior52.6 in.
Height (Metric)154.5 cm
Height (Metric) Interior133.5 cm
Depth (English) Interior21.5 in.
Depth (Metric) Interior54.5 cm
Electrical Requirements230 V 50/60 Hz
Voltage230 V
Wattage3000 w
Amperage13 A
Frequency50/60 Hz
Unit SizeEach
Showing 10 of 12Show more
Catalog NumberSpecificationsUnit SizeCapacity (Metric)TypeMaterialPlug TypePrice (USD)
51028144Full specifications
Each168 LGravity ConvectionCoated SteelCountry-specificRequest A Quote
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Description51028144
Capacity (Metric)168 L
Temperature (Metric)330°C
TypeGravity Convection
MaterialCoated Steel
No. of Shelves2 supplied/19 max.
Plug TypeCountry-specific
Temperature Stability±2.5°C
Dimensions (D x W x H) Interior22.2 x 17.2 x 26.8 in. (56.4 x 43.8 x 68 cm)
Dimensions (L x W x H)29.1 x 25.2 x 36.2 in. (73.8 x 64 x 92 cm)
Length (English)29.1 in.
Length (Metric)73.8 cm
Width (English)25.2 in.
Width (English) Interior17.2 in.
Width (Metric)64 cm
Width (Metric) Interior43.8 cm
Height (English)36.2 in.
Height (English) Interior26.8 in.
Height (Metric)92 cm
Height (Metric) Interior68 cm
Depth (English) Interior22.2 in.
Depth (Metric) Interior56.4 cm
Electrical Requirements230 V 50/60 Hz
Voltage230 V
Wattage3100 w
Amperage13.5 A
Frequency50/60 Hz
Unit SizeEach
51028142Full specifications
Each61 LGravity ConvectionCoated SteelCountry-specificRequest A Quote
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Description51028142
Capacity (Metric)61 L
Temperature (Metric)330°C
TypeGravity Convection
MaterialCoated Steel
No. of Shelves2 supplied/13 max.
Plug TypeCountry-specific
Temperature Stability±2.5°C
Dimensions (D x W x H) Interior15.3 x 12.9 x 18.9 in. (38.9 x 32.8 x 48 cm)
Dimensions (L x W x H)22.2 x 20.9 x 28.3 in. (56.5 x 53 x 72 cm)
Length (English)22.2 in.
Length (Metric)56.5 cm
Width (English)20.9 in.
Width (English) Interior12.9 in.
Width (Metric)53 cm
Width (Metric) Interior32.8 cm
Height (English)28.3 in.
Height (English) Interior18.9 in.
Height (Metric)72 cm
Height (Metric) Interior48 cm
Depth (English) Interior15.3 in.
Depth (Metric) Interior38.9 cm
Electrical Requirements230 V 50/60 Hz
Voltage230 V
Wattage1800 w
Amperage7.9 A
Frequency50/60 Hz
Unit SizeEach
51028151Full specifications
Each62 LMechanical Convection OvenCoated SteelCountry-specificRequest A Quote
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Description51028151
Capacity (Metric)62 L
Temperature (Metric)330°C
TypeMechanical Convection Oven
MaterialCoated Steel
No. of Shelves2 supplied/13 max.
Plug TypeCountry-specific
Temperature Stability±1.8°C
Dimensions (D x W x H) Interior13.5 x 13.9 x 20 in. (34.3 x 35.4 x 50.8 cm)
Dimensions (L x W x H)22.2 x 20.9 x 28.3 in. (56.5 x 53 x 72 cm)
Length (English)22.2 in.
Length (Metric)56.5 cm
Width (English)20.9 in.
Width (English) Interior13.9 in.
Width (Metric)53 cm
Width (Metric) Interior35.4 cm
Height (English)28.3 in.
Height (English) Interior20 in.
Height (Metric)72 cm
Height (Metric) Interior50.8 cm
Depth (English) Interior13.5 in.
Depth (Metric) Interior34.3 cm
Electrical Requirements230 V 50/60 Hz
Voltage230 V
Wattage1400 w
Amperage6.1 A
Frequency50/60 Hz
Unit SizeEach
51028152Full specifications
Each97 LMechanical Convection OvenCoated SteelCountry-specificRequest A Quote
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Description51028152
Capacity (Metric)97 L
Temperature (Metric)330°C
TypeMechanical Convection Oven
MaterialCoated Steel
No. of Shelves2 supplied/16 max.
Plug TypeCountry-specific
Temperature Stability±1.3°C
Dimensions (D x W x H) Interior13.5 x 18.3 x 23.9 in. (34.3 x 46.4 x 60.8 cm)
Dimensions (L x W x H)22.2 x 25.2 x 32.3 in. (56.5 x 64 x 82 cm)
Length (English)22.2 in.
Length (Metric)56.5 cm
Width (English)25.2 in.
Width (English) Interior18.3 in.
Width (Metric)64 cm
Width (Metric) Interior46.4 cm
Height (English)32.3 in.
Height (English) Interior23.9 in.
Height (Metric)82 cm
Height (Metric) Interior60.8 cm
Depth (English) Interior13.5 in.
Depth (Metric) Interior34.3 cm
Electrical Requirements230 V 50/60 Hz
Voltage230 V
Wattage3060 w
Amperage13.3 A
Frequency50/60 Hz
Unit SizeEach
51028153Full specifications
Each170 LMechanical Convection OvenCoated SteelCountry-specificRequest A Quote
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Description51028153
Capacity (Metric)170 L
Temperature (Metric)330°C
TypeMechanical Convection Oven
MaterialCoated Steel
No. of Shelves2 supplied/19 max.
Plug TypeCountry-specific
Temperature Stability±1.8°C
Dimensions (D x W x H) Interior20.4 x 18.3 x 27.9 in. (51.8 x 46.4 x 70.8 cm)
Dimensions (L x W x H)29.1 x 25.2 x 36.2 in. (73.8 x 64 x 92 cm)
Length (English)29.1 in.
Length (Metric)73.8 cm
Width (English)25.2 in.
Width (English) Interior18.3 in.
Width (Metric)64 cm
Width (Metric) Interior46.4 cm
Height (English)36.2 in.
Height (English) Interior27.9 in.
Height (Metric)92 cm
Height (Metric) Interior70.8 cm
Depth (English) Interior20.4 in.
Depth (Metric) Interior51.8 cm
Electrical Requirements230 V 50/60 Hz
Voltage230 V
Wattage3060 w
Amperage13.3 A
Frequency50/60 Hz
Unit SizeEach
51028709Full specifications
Each61 LGravity ConvectionStainless SteelCountry-specificRequest A Quote
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Description51028709
Capacity (Metric)61 L
Temperature (Metric)330°C
TypeGravity Convection
MaterialStainless Steel
No. of Shelves2 supplied/13 max.
Plug TypeCountry-specific
Temperature Stability±2.5°C
Dimensions (D x W x H) Interior15.3 x 12.9 x 18.9 in. (38.9 x 32.8 x 48 cm)
Dimensions (L x W x H)22.2 x 20.9 x 28.3 in. (56.5 x 53 x 72 cm)
Length (English)22.2 in.
Length (Metric)56.5 cm
Width (English)20.9 in.
Width (English) Interior12.9 in.
Width (Metric)53 cm
Width (Metric) Interior32.8 cm
Height (English)28.3 in.
Height (English) Interior18.9 in.
Height (Metric)72 cm
Height (Metric) Interior48 cm
Depth (English) Interior15.3 in.
Depth (Metric) Interior38.9 cm
Electrical Requirements230 V 50/60 Hz
Voltage230 V
Wattage1800 w
Amperage7.9 A
Frequency50/60 Hz
Unit SizeEach
51028713Full specifications
Each62 LMechanical Convection OvenStainless SteelCountry-specificRequest A Quote
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Description51028713
Capacity (Metric)62 L
Temperature (Metric)330°C
TypeMechanical Convection Oven
MaterialStainless Steel
No. of Shelves2 supplied/13 max.
Plug TypeCountry-specific
Temperature Stability±1.8°C
Dimensions (D x W x H) Interior13.5 x 13.9 x 20 in. (34.3 x 35.4 x 50.8 cm)
Dimensions (L x W x H)22.2 x 20.9 x 28.3 in. (56.5 x 53 x 72 cm)
Length (English)22.2 in.
Length (Metric)56.5 cm
Width (English)20.9 in.
Width (English) Interior13.9 in.
Width (Metric)53 cm
Width (Metric) Interior35.4 cm
Height (English)28.3 in.
Height (English) Interior20 in.
Height (Metric)72 cm
Height (Metric) Interior50.8 cm
Depth (English) Interior13.5 in.
Depth (Metric) Interior34.3 cm
Electrical Requirements230 V 50/60 Hz
Voltage230 V
Wattage1400 w
Amperage6.1 A
Frequency50/60 Hz
Unit SizeEach
51028715Full specifications
Each97 LMechanical Convection OvenStainless SteelCountry-specificRequest A Quote
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Description51028715
Capacity (Metric)97 L
Temperature (Metric)330°C
TypeMechanical Convection Oven
MaterialStainless Steel
No. of Shelves2 supplied/16 max.
Plug TypeCountry-specific
Temperature Stability±1.3°C
Dimensions (D x W x H) Interior13.5 x 18.3 x 23.9 in. (34.3 x 46.4 x 60.8 cm)
Dimensions (L x W x H)22.2 x 25.2 x 32.3 in. (56.5 x 64 x 82 cm)
Length (English)22.2 in.
Length (Metric)56.5 cm
Width (English)25.2 in.
Width (English) Interior18.3 in.
Width (Metric)64 cm
Width (Metric) Interior46.4 cm
Height (English)32.3 in.
Height (English) Interior23.9 in.
Height (Metric)82 cm
Height (Metric) Interior60.8 cm
Depth (English) Interior13.5 in.
Depth (Metric) Interior34.3 cm
Electrical Requirements230 V 50/60 Hz
Voltage230 V
Wattage3060 w
Amperage13.3 A
Frequency50/60 Hz
Unit SizeEach
51028716Full specifications
Each170 LMechanical Convection OvenStainless SteelCountry-specificRequest A Quote
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Description51028716
Capacity (Metric)170 L
Temperature (Metric)330°C
TypeMechanical Convection Oven
MaterialStainless Steel
No. of Shelves2 supplied/19 max.
Plug TypeCountry-specific
Temperature Stability±1.8°C
Dimensions (D x W x H) Interior20.4 x 18.3 x 27.9 in. (51.8 x 46.4 x 70.8 cm)
Dimensions (L x W x H)29.1 x 25.2 x 36.2 in. (73.8 x 64 x 92 cm)
Length (English)29.1 in.
Length (Metric)73.8 cm
Width (English)25.2 in.
Width (English) Interior18.3 in.
Width (Metric)64 cm
Width (Metric) Interior46.4 cm
Height (English)36.2 in.
Height (English) Interior27.9 in.
Height (Metric)92 cm
Height (Metric) Interior70.8 cm
Depth (English) Interior20.4 in.
Depth (Metric) Interior51.8 cm
Electrical Requirements230 V 50/60 Hz
Voltage230 V
Wattage3060 w
Amperage13.3 A
Frequency50/60 Hz
Unit SizeEach
51029332Full specifications
Each396 LMechanical Convection OvenStainless SteelCountry-specificRequest A Quote
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Description51029332
Capacity (Metric)396 L
Temperature (Metric)330°C
TypeMechanical Convection Oven
MaterialStainless Steel
No. of Shelves2 supplied/39 max.
Plug TypeCountry-specific
Temperature Stability±2.1°C
Dimensions (D x W x H) Interior21.5 x 21.4 x 52.6 in. (54.5 x 54.4 x 133.5 cm)
Dimensions (L x W x H)30.3 x 30.6 x 60.8 in. (77 x 77.8 x 154.5 cm)
Length (English)30.3 in.
Length (Metric)77 cm
Width (English)30.6 in.
Width (English) Interior21.4 in.
Width (Metric)77.8 cm
Width (Metric) Interior54.4 cm
Height (English)60.8 in.
Height (English) Interior52.6 in.
Height (Metric)154.5 cm
Height (Metric) Interior133.5 cm
Depth (English) Interior21.5 in.
Depth (Metric) Interior54.5 cm
Electrical Requirements230 V 50/60 Hz
Voltage230 V
Wattage3000 w
Amperage13 A
Frequency50/60 Hz
Unit SizeEach
Showing 10 of 12Show more
Features:
  • Broad temperature range—up to 330°C
  • Adjustable fan speed for application-related airflow in mechanical convection models (e.g. drying of powders)
  • High speed for expedited drying and heating, quick recovery, and optimal temperature stability and uniformity
  • Programmable controller for temperature ramps and dwells
  • Up to 10 programs can be saved, each with 10 discrete steps
  • Features electronically controlled fan speed and damper position
  • Programs can be repeated automatically
  • Access port allows the introduction of sensors for independent data monitoring
  • Simple calibration routine ensures temperature accuracy over time
  • Boost function enables rapid heating up—no need to operate the oven 24/7 (not available in large capacity units)
Ordering Information:
Availability may vary by country.
Warranty: 2 year warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.