Nalgene™ BioTransport Carrier Handles
Nalgene™ BioTransport Carrier Handles
Thermo Scientific™
Nalgene™ BioTransport Carrier Handles
Securely hold a fully loaded carrier with Thermo Scientific™ Nalgene™ BioTransport Carrier Handles, which fold conveniently to the side for storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionUnit Size
7136-0001Bio Transport Carrier HandleCase of 8
7136-0001PK
also known as 7136-0001
Bio Transport Carrier HandleEach
Catalog number 7136-0001
Price (USD)
-
Request A Quote
Description:
Bio Transport Carrier Handle
Unit Size:
Case of 8
Ask our AI about this Product
Securely hold a fully loaded carrier with Thermo Scientific™ Nalgene™ BioTransport Carrier Handles, which fold conveniently to the side for storage. These handles fit the BioTransport carrier for easy one-handed carrying.
  • Simple to put on and take off
  • Autoclavable stainless steel
  • Includes: Handle only. Biotransport Carrier sold separately, Cat. No. 7135-0001
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.
Specifications
DescriptionBio Transport Carrier Handle
AutoclavableAutoclavable
Product TypeBioTransport Carrier Handle
Unit SizeCase of 8
Lab Safety and Organization
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Los clientes que vieron este artículo también vieron
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection