MagniSort™ Mouse CD4 Positive Selection Kit
Product Image
Invitrogen™

MagniSort™ Mouse CD4 Positive Selection Kit

The MagniSort™ Mouse CD4 Positive Selection Kit is designed for the magnetic separation of CD4+ cells by positive selection. ItRead more
Promo ImagePromotion
Promo code:RFLOW25
Get more, spend less on every flow cytometry run
Fuel discovery without breaking your budget. Save up to 33% on flow cytometry essentials—compensation beads, buffers, assays & more. Use code RFLOW25 at checkout. The more you buy, the more you save. Limited time only!Learn More
Have Questions?
Catalog NumberQuantity
8802-6841-74Promo Image200 tests
Catalog number 8802-6841-74
Price (USD)
976.00
Each
In stock
Add to cart
Quantity:
200 tests
Recurring order eligible. Learn more »
Price (USD)
976.00
Each
Add to cart
Ask our AI about this Product
The MagniSort™ Mouse CD4 Positive Selection Kit is designed for the magnetic separation of CD4+ cells by positive selection. It has been optimized for the isolation of CD4+ cells from mouse spleens or lymph nodes utilizing a biotinylated Anti-Mouse CD4 antibody and streptavidin-coated magnetic beads. CD4+ cells are bound by antibody and then magnetic beads. When placed in a magnetic field, the undesired cells can be separated from CD4+ cells by decanting.

After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD4, clones RM4-5 or GK1.5.

Reported Application
Magnetic Cell Separation

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeCD4+
No. of Tests200 tests
Quantity200 tests
Target SpeciesMouse
Product TypePositive Selection Kit
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2913935Certificate of AnalysisMay 07, 20248802-6841-74
2829454Certificate of AnalysisDec 21, 20238802-6841-74
2618909Certificate of AnalysisMar 24, 20238802-6841-74
2528509Certificate of AnalysisSep 02, 20228802-6841-74
2350776Certificate of AnalysisJul 08, 20218802-6841-74
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection