GeneChip™ Human Genome U133A 2.0 Array
GeneChip™ Human Genome U133A 2.0 Array
Applied Biosystems™
GeneChip™ Human Genome U133A 2.0 Array
The GeneChip™ Human Genome U133A 2.0 Array is a single array representing 14,500 well-characterized human genes that can be usedRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNumber of Arrays
90046930 arrays
9004712 arrays
9004686 arrays
Catalog number 900469
Price (USD)
-
Number of Arrays:
30 arrays
Ask our AI about this Product
The GeneChip™ Human Genome U133A 2.0 Array is a single array representing 14,500 well-characterized human genes that can be used to explore human biology and disease processes.
The GeneChip™ Human Genome U133A 2.0 Array is a single array representing 14,500 well-characterized human genes that can be used to explore human biology and disease processes. New design and reduced feature size mean that you can use smaller sample volumes than the previous HG™U133A Array without compromising performance.
•Provides coverage of well-substantiated genes in the transcribed human genome on a single array
•Analyzes the expression level of 18,400 transcripts and variants, including 14,500 well-characterized human genes
•Comprised of more than 22,000 probe sets and 500,000 distinct oligonucleotide features
•Use the Power of the Probe Set and get multiple independent measurements for each transcript that deliver the greatest accuracy and reproducibility of any microarray platform
•All probe sets represented on the GeneChip™ Human Genome U133A Array are identically replicated on the GeneChip Human Genome U133A 2.0 Array
Array Profile
Sequences used in the design of the array were selected from GenBank™, dbEST, and RefSeq. The sequence clusters were created from the UniGene database (Build 133, April 20, 2001) and then were refined by analysis and comparison with a number of other publicly available databases including the Washington University EST trace repository and the University of California, Santa Cruz Golden-Path human genome database (April 2001 release).
Instrument and Software Requirements
•GeneChip™ Scanner 3000, enabled for High-Resolution Scanning*
•GeneChip™ Command Console™ Software (AGCC) including the GeneChip™ Scanner 3000 High-Resolution Scanning Patch
*GeneChip Scanner 3000 High-Resolution Update is standard on all instruments shipped starting in September 2003 with serial number series 502. Previous versions, serial number series 501, will require the 00-0110 GeneChip Scanner 3000 High-Resolution Update to be installed.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
TypeHuman Genome U133A 2.0 Array
ArrayTranscriptome Profiling
Number of Arrays30 arrays
FormatArray Cartridge
SpeciesHuman
Product LineGeneChip™
Quantity30 arrays
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Customers who viewed this item also viewed
Frequently asked questions (FAQs)
The intended use of the 100 probe sets is to allow users to normalize data across the chip. These 100 probe sets are considered normalization controls. They were found to have consistent levels of expression across multiple tissue types and treatments.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Occasionally, it is not possible to select either a unique probe set or a probe set with all probes common among multiple transcripts ("_s_at" ). In such cases, similarity criteria are suspended, and the resulting probe set name is appended with the "_x_at" extension. These probe sets contain some probes that are identical, or highly similar, to unrelated sequences. These probes may cross-hybridize in an unpredictable manner with sequences other than the main target. Data generated from these probe sets should be interpreted with caution, due to the likelihood that some of the signal is from transcripts other than the one being intentionally measured.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
The primary goal in probe set selection is to select a probe set unique to a single transcript or common among a small set of similar transcript variants. A probe set name is appended with the "_s_at" extension when all the probes exactly match multiple transcripts. The probe set selection process generally favors probe sets measuring fewer transcripts. Probe sets with common probes among multiple transcripts (the "_s_at" probe sets), are frequent and are to be expected, due to alternative polyadenylation and alternative splicing. In most cases, "_s_at" probe sets represent transcripts from the same gene, but the same probe set can sometimes also represent transcripts from homologous genes. One transcript may be represented by both a unique and an "_s_at" probe set when the transcript variation is sufficient.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
There are two ways to export the CEL file from GCOS v1.4 as a text file.
1. In GCOS 1.4 use GCOS Manager to navigate to the CEL file in the process database. Highlight the CEL file and right click to export the data from the process tab. You may want to change the data location where the export will be stored before you start.
2. In GCOS 1.4 use the find feature in GCOS Manager to find the CEL files and export those selected as a text file. This approach allows export of more than one experiment at a time.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Command Console cannot generate a comparison analysis between different array types.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Share catalog number, name or link
1x1 image pixel for data collection