Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | TMT10plex™ Isobaric Label Reagent Set, 8 x 0.2 mg |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Isobaric Label Reagent Set |
Product Line | TMT10plex™ |
Label or Dye | Tandem Mass Tag™ (TMT) Labels |
Shipping Condition | Dry Ice |
Starting Material | Peptides |
Detection Method | Mass Spectrometry |
Workflow Step | Peptide quantitation |
Quantity | 8 x 0.2 mg |
Unit Size | 8 x 0.2 mg (per tag) |
Description | TMT10plex™ Isobaric Label Reagent Set, 1 x 0.8 mg |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Isobaric Label Reagent Set |
Product Line | TMT10plex™ |
Label or Dye | Tandem Mass Tag™ (TMT) Labels |
Starting Material | Cell Lysate |
Final Product Type | Protein PEPTIDE |
Detection Method | Mass Spectrometry |
Workflow Step | Protein Labeling |
Quantity | 10 Reactions |
Unit Size | 1 x 0.8 mg (per tag) |
Description | TMT10plex™ Isobaric Label Reagent Set, 3 x 0.8 mg |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Isobaric Label Reagent Set |
Product Line | TMT10plex™ |
Label or Dye | Tandem Mass Tag™ (TMT) Labels |
Starting Material | Cell Lysate |
Final Product Type | Protein |
Detection Method | Mass Spectrometry |
Workflow Step | Protein Labeling |
Quantity | 30 Reactions |
Unit Size | 3 x 0.8 mg (per tag) |
Description | TMT10plex™ Isobaric Label Reagent Set, 1 x 5 mg |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Isobaric Label Reagent Set |
Product Line | TMT10plex™ |
Label or Dye | Tandem Mass Tag™ (TMT) Labels |
Starting Material | Cell Lysate |
Final Product Type | Protein PEPTIDE |
Detection Method | Mass Spectrometry |
Workflow Step | Protein Labeling |
Quantity | 60 Reactions |
Unit Size | 1 x 5 mg (per tag) |
Description | TMT10plex™ Isobaric Mass Tagging Kit |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Isobaric Mass Tag Labeling Kit |
Product Line | TMT10plex™ |
Label or Dye | Tandem Mass Tag™ (TMT) Labels |
Starting Material | Cell Lysate |
Final Product Type | Protein |
Detection Method | Mass Spectrometry |
Workflow Step | Protein Labeling |
Quantity | 30 Reactions |
Unit Size | 30 reactions |
Description | TMT™ 10plex Label Reagents in stabilized solution, 96-well plate, 8 x 40 μg |
Formulation | Liquid in stabilized DMSO solution |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Isobaric Label Reagent Set |
Product Line | TMT10plex™ |
Label or Dye | Tandem Mass Tag™ (TMT) Labels |
Starting Material | Cell Lysate |
Final Product Type | Protein PEPTIDE |
Detection Method | Mass Spectrometry |
Workflow Step | Protein Labeling |
Quantity | 80 Reactions |
Form | Liquid |
Unit Size | 8 x 40 μg (per tag) |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
90309 | 8 x 0.2 mg (per tag) | TMT10plex™ Isobaric Label Reagent Set, 8 x 0.2 mg | 4,535.65 Online price 4,995.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
90110 | 1 x 0.8 mg (per tag) | TMT10plex™ Isobaric Label Reagent Set, 1 x 0.8 mg | 1,243.65 Online price 1,278.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
90111 | 3 x 0.8 mg (per tag) | TMT10plex™ Isobaric Label Reagent Set, 3 x 0.8 mg | 3,068.65 Online price 3,440.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
90406 | 1 x 5 mg (per tag) | TMT10plex™ Isobaric Label Reagent Set, 1 x 5 mg | 5,595.65 Online price 5,940.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
90113 | 30 reactions | TMT10plex™ Isobaric Mass Tagging Kit | 3,397.65 Online price 3,725.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A58332 | 8 x 40 μg (per tag) | TMT™ 10plex Label Reagents in stabilized solution, 96-well plate, 8 x 40 μg | 600.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The Tandem Mass Tag (TMT) Isobaric Label reagents and kits are specifically designed to enable identification and quantitation of proteins in different samples using tandem mass spectrometry. The TMT10plex label reagents share an identical structure with TMTzero, TMTduplex, and TMTsixplex reagents, but contain different numbers and combinations of 13C and 15N isotopes in the mass reporter. The different isotopes result in a 10-plex set of tags that have mass differences in the reporter that can be detected using high resolution Orbitrap MS instruments.
Advantages of the TMT10plex label reagents include increased multiplex relative quantitation, increased sample throughput, and fewer missing quantitative values among samples. TMT10plex label reagents are ideal for analysis of multiple protein samples from inhibitor dose response experiments, time course experiments, or improving statistics on biological replicates.
Features of TMT10plex Isobaric Label reagent sets and mass tagging kits
• Powerful—multiplexed MS analysis increases sample throughput and enables relative quantitation of up to ten different samples derived from cells, tissues, or biological fluids
• Consistent—identical reagent structure and performance among TMTzero, TMTduplex, TMTsixplex, and TMT10plex reagents allows efficient transition from method development to multiplex quantitation, enabling biomarker discovery research across platforms and datasets
• Robust—increased multiplex capability results in fewer missing quantitative values among samples and higher confidence among replicates
• Efficient—amine-reactive, NHS-ester-activated reagents ensure efficient labeling of all peptides regardless of protein sequence or proteolytic enzyme specificity
• Compatible—optimized for use with high resolution Orbitrap MS/MS platforms, with data analysis fully supported by Proteome Discoverer 2.3.
Applications:
• Protein identification and quantitation from multiple samples of cells, tissue, or biological fluids
• Protein expression profiling of normal vs. disease states or control vs. treated
• Quantitative analysis of proteins for which no antibodies are available
• Identification and quantitation of membrane and post-translationally modified proteins
• Identification and quantification of hundreds to thousands of proteins in a single experiment
TMT Reagents are provided as standalone sets, a 96-well format, or in optimized kit formats containing all necessary reagents and controls for maximum flexibility, convenience, and reliability. When combined with the industry-leading, high resolution Thermo Scientific Orbitrap instruments and software, TMT10plex reagents provide integrated total solutions for quantitative protein expression analysis.