Search Thermo Fisher Scientific
Ensure accurate, precise pipetting. Finntip™ Specific Pipette Tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
Catalog Number | Volume (Metric) | Sterility | Fit for Purpose | Color |
---|---|---|---|---|
9401115 | 1000 μL | Non-sterile | Certified Quality | Blue |
9400300 | 10 μL | Non-sterile | Certified Quality | Pink |
9400303 | 10 μL | Sterile | Sterile & PCR Ready | Pink |
9400310 | 10 μL | Non-sterile | Certified Quality | Pink |
9400327 | 10 μL | Non-sterile | Certified Quality | Pink |
9401030 | 1000 μL | Non-sterile | Certified Quality | Blue |
9401110 | 1000 μL | Non-sterile | Certified Quality | Blue |
9401113 | 1000 μL | Sterile | Sterile & PCR Ready | Blue |
9401250 | 300 μL | Non-sterile | Certified Quality | Orange |
9401253 | 300 μL | Sterile | Sterile & PCR Ready | Orange |
9401255 | 300 μL | Non-sterile | Certified Quality | Orange |
9401260 | 300 μL | Non-sterile | Certified Quality | Orange |
9402030 | 5 mL | Non-sterile | Certified Quality | Green |
9402070 | 5 mL | Non-sterile | Certified Quality | Green |
9402073 | 5 mL | Sterile | Sterile & PCR Ready | Green |
9402171 | 10 mL | Non-sterile | Certified Quality | Red |
9402180 | 10 mL | Non-sterile | Certified Quality | Red |
9402183 | 10 mL | Sterile | Sterile & PCR Ready | Red |
9400360 | 50 μL | Non-sterile | Certified Quality | Turquoise |
9400370 | 50 μL | Non-sterile | Certified Quality | Turquoise |
9400373 | 50 μL | Sterile | Sterile & PCR Ready | Turquoise |
Key Features:
Proven quality
Versatile Tip Range
Immediate traceability
Note:
Availability may vary by country.
Gain confidence that the products you buy are ideally suited to your application.
Share catalog number, name or link