ClipTip™ Filtered Pipette Tips
ClipTip™ Filtered Pipette Tips
ClipTip™ Filtered Pipette Tips
ClipTip™ Filtered Pipette Tips
Thermo Scientific™

ClipTip™ Filtered Pipette Tips

Ensure a secure seal for every sample. A unique, innovative interlocking technology ensures a complete seal on every channel with minimal tip attachment and ejection force.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Format
94420613300 μLRack
9442004312.5 μLRack
9442021820 μLReload Deck
9442025850 μLReload Deck
94420318200 μLReload Deck
944207181000 μLReload Deck
944208131250 μLRack
944208181250 μLReload Deck
9442021320 μLRack
9442025350 μLRack
94420313200 μLRack
94420513300 μLRack
944207131000 μLRack
9442006312.5 μLRack
Catalog number 94420613
Price (EUR)
70,40
Pack of 768
-
Add to cart
Volume (Metric):
300 μL
Format:
Rack
Price (EUR)
70,40
Pack of 768
Add to cart
Ask our AI about this Product
Ensure a secure seal for every sample. Thermo Scientific™ ClipTip™ Pipette Tips provide a secure seal for every sample with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force. Achieve newfound confidence knowing that once attached, your tips are locked firmly in place, and will not loosen or fall off regardless of application pressure. The innovative three interlocking clip design ensures the tip is held securely on the F1-ClipTip and E1-ClipTip pipettes until — and only until — it is released.

Key Features:

  • ClipTip pipette tips feature 'clips' that lock tips in place and form a complete seal
  • Each time ClipTip pipette tips are on, they are sealed and will not loosen regardless of application pressure
  • ClipTip pipette tips are clipped securely on the F1-ClipTip and E1-ClipTip pipettes with a light touch eliminating the frustration of banging tips on your pipette
  • Filtered ClipTip pipette tips are ideal for sensitive assays, such as PCR, and with critical samples to prevent cross-contaminations

Low retention

  • Low retention technology minimizes liquid retention, and enable maximium sample recovery. Available in ClipTip 20-1250 96-format tips.


Proven quality

  • Certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination


Environmentally responsible

  • ClipTip reload system uses recycled components and 30% less plastic


Versatile Tip Range

  • 96-format volumes: 12.5, 20, 50, 200, 300, 1000 and 1250μL
  • 384-format volumes: 12.5, 30 and 125μL
  • Extended length tips available for volumes 12.5 and 300 μl allowing for better reach with specific labware consumables


Immediate traceability

  • Each tip insert has ink-jetted lot and volume information; the convenient color key makes it easy to match the color code of the F1-ClipTip and E1-ClipTip pipette with the corresponding ClipTip pipette tip rack insert


Note:

Availability may vary by country.

Specifications
DescriptionClipTip 300 Ext, Filtered, Sterile, Rack
Length (English)3.94 in
Length (Metric)10 cm
Packaging8 x 96 tips/rack
AutoclavableNot Autoclavable
Color CodeOrange
FilteredFiltered
Fit for PurposeSterile & PCR Ready
For Use With (Equipment)Thermo Scientific E1-ClipTip, Thermo Scientific F1-ClipTip
FormatRack
Product LineClipTip
SterilitySterile
Tip StyleLow Retention, Extended Length
Volume (Metric)300 μL
Unit SizePack of 768
cliptip-technology

Application notes

  Learn what makes the new ClipTip technology a truly different experience

web-banner-9-8-22-278x123

Fit for Purpose designations

Gain confidence that the products you buy are ideally suited to your application. 

Learn more ›

Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
25163C0Certificate of ComplianceJun 27, 202594420613
25170C0Certificate of ComplianceJun 27, 202594420213
25147C0Certificate of ComplianceJun 18, 202594420813
25144C0Certificate of ComplianceJun 18, 202594420813
25122C0Certificate of ComplianceJun 18, 202594420813
5 results displayed, search above for a specific certificate

Share catalog number, name or link

1x1 image pixel for data collection