AcroMetrix Multi-Analyte SARS-CoV-2, Flu A/B, RSV A/B Control (RUO)
AcroMetrix Multi-Analyte SARS-CoV-2, Flu A/B, RSV A/B Control (RUO)
Thermo Scientific™

AcroMetrix Multi-Analyte SARS-CoV-2, Flu A/B, RSV A/B Control (RUO)

Validate and monitor the performance of molecular diagnostic tests for detecting COVID-19, Flu A/B and RSV A/B using the Thermo Scientific™ AcroMetrix™ Multi-Analyte SARS-CoV-2, Flu A/B, RSV A/B Control (RUO).
製品番号(カタログ番号)数量
9545161 Kit
製品番号(カタログ番号) 954516
価格(JPY)
-
見積もりを依頼する
数量:
1 Kit
Ask our AI about this Product
Validate and monitor the performance of molecular diagnostic tests for detecting COVID-19, Flu A/B and RSV A/B using the AcroMetrix™ Multi-Analyte SARS-CoV-2 Control, Flu A/B, RSV A/B Control (RUO). This multiplex control can be used as a complete assay control starting from extraction to amplification and finally detection of COVID-19, Flu A/B and RSV A/B.
  • Inactivated SARS-CoV-2, Influenza A/B and Respiratory Syncytial Virus A/B in proprietary viral transport media
  • Full process control mimics a clinical sample
  • Multiplexed quantitative positive control
  • Kit contains 5 vials per kit x 1.5 mL per vial
研究用にのみご使用ください。診断用には対応していません。
For Research Use Only. Not for use in diagnostic procedures.
仕様
概要AcroMetrix Multi-Analyte SARS-CoV-2, Flu A/B, RSV A/B Control (RUO)
パッケージOne kit contains 5 vials per kit x 1.5 mL per vial
数量1 Kit
Unit SizeEach
組成および保存条件
-20°C
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

ドキュメントおよびダウンロード

証明書

Safety Data Sheets

こちらの製品には複数の製品番号が設定されています。