Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Format | 96-well Comb |
Capacity | 96 Samples |
Description | KingFisher 96 tip comb for KingFisher PCR magnets, 80 pcs (for Flex only) |
For Use With (Equipment) | KingFisher Flex only |
Quantity | 80/Cs. |
Unit Size | Case of 80 |
Format | 96-well Comb |
Capacity | 96 Samples |
Description | KingFisher Apex 96 PCR tip comb (for Apex only) |
Disposable | Single-use |
For Use With (Equipment) | KingFisher Apex only |
Quantity | 50/Cs. |
Unit Size | Case of 50 |
Format | 96 Deep-well Plate |
Capacity | 96 Samples |
Description | KingFisher 96 microplate (200μL) (for Flex and Presto) *Note: Only works with KingFisher™ Flex 96 Heating Block (Cat. No. 24075420). Not to be used with KingFisher™ Flex 96 Deep-Well Heating Block (Cat. No. 24075430). |
For Use With (Equipment) | KingFisher Flex and Presto |
Unit Size | Case of 48 |
Format | 96-well Comb |
Capacity | 96 Samples |
Description | KingFisher 96 tip comb for KingFisher magnets, 100 pcs (for Flex) |
For Use With (Equipment) | KingFisher Flex |
Unit Size | Case of 100 |
Description | KingFisher 96 Tip Comb, Barcoded |
For Use With (Equipment) | KingFisher Apex |
Quantity | 100/Cs. |
Unit Size | Case of 100 |
Capacity | 200 μL |
Description | KingFisher 96 Plate (200 μL), Barcoded |
For Use With (Equipment) | KingFisher Apex |
Quantity | 48/Cs. |
Unit Size | Case of 48 |
Catalog Number | Specifications | Unit Size | Description | For Use With (Equipment) | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
97002514 | Case of 80 | KingFisher 96 tip comb for KingFisher PCR magnets, 80 pcs (for Flex only) | KingFisher Flex only | 480.65 Online price 500.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
97002560 | Case of 50 | KingFisher Apex 96 PCR tip comb (for Apex only) | KingFisher Apex only | 351.65 Online price 366.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
97002540 | Case of 48 | KingFisher 96 microplate (200μL) (for Flex and Presto) *Note: Only works with KingFisher™ Flex 96 Heating Block (Cat. No. 24075420). Not to be used with KingFisher™ Flex 96 Deep-Well Heating Block (Cat. No. 24075430). | KingFisher Flex and Presto | 184.65 Online price 230.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
97002524 | Case of 100 | KingFisher 96 tip comb for KingFisher magnets, 100 pcs (for Flex) | KingFisher Flex | 589.65 Online price 614.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
97002524B | Case of 100 | KingFisher 96 Tip Comb, Barcoded | KingFisher Apex | 543.65 Online price 632.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
97002540B | Case of 48 | KingFisher 96 Plate (200 μL), Barcoded | KingFisher Apex | 222.65 Online price 232.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|