Gibco™

TrypLE™ Select Enzyme (10X), no phenol red

Catalog number: A1217703
Gibco™

TrypLE™ Select Enzyme (10X), no phenol red

Catalog number: A1217703
Catalog Number
A1217703
Unit Size
20 x 100 mL
Price (MXN)
Availability
***
Quantity
-
Catalog Number
Unit Size
100 mL
Price (MXN)
Price: 4,354.05
Your price:
Availability
***
Quantity
Catalog Number
Unit Size
500 mL
Price (MXN)
Price: 17,856.23
Your price:
Availability
***
Quantity
Catalog NumberUnit SizePrice (MXN)AvailabilityQuantity
A121770320 x 100 mLContact Us ›***-
A1217701100 mL
Price: 4,354.05
Your price:
***
A1217702500 mL
Price: 17,856.23
Your price:
***
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
TrypLE™ Select is an animal origin free, recombinant enzyme for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A549, primary human keratinocytes, and embryonic stem cells. We offer a variety of TrypLE™ formulations for a range of cell culture applications.

Compared to trypsin and other dissociation reagents, Gibco™ TrypLE™ Select is:

Gentle on cells—maintains cell health for reproducible results
Room-temperature stable—convenient to use
Easy to use—substitutes directly into existing protocols, with no inactivation required
Animal origin free (AOF)—pure, consistent, and effective

This TrypLE™ Select is modified as follows:

With
• EDTA

Without
• Phenol Red

The formulation of TrypLE™ solutions is available, although we keep the enzyme concentration confidential.

Gentle on cells
TrypLE™ Select, like trypsin, cleaves peptide bonds on the C-terminal sides of lysine and arginine. However, TrypLE™ Select's exceptional purity increases specificity due to the action of a single enzyme. This reduces damage caused by cleavage from multiple enzymes in trypsin and other extracts.

Room-temperature stable
TrypLE™ Select is room-temperature stable and ready to use. TrypLE™ Select cell dissociation reagents remain stable for 24 months at room temperature or 2–8°C, making storage and handling easy and convenient.

Easy to use
TrypLE™ Select can be substituted for trypsin in existing protocols. In addition, dilution alone inactivates TrypLE™ Select, avoiding the need for trypsin inhibitors, such as FBS.

Animal Origin Free (AOF)
Unlike porcine trypsin and other enzymes, TrypLE™ Select is free of animal-derived components, and is formulated on dedicated animal origin-free equipment.

Product Intended Use
For Research Use or Further Manufacturing. Not for use in diagnostic procedures. Customers using Gibco™ TrypLE™ Select in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

cGMP Manufacturing and Quality System
Gibco™ TrypLE™ Select is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Concentrated forms of TrypLE™ can be aseptically diluted to 1x using a balanced salt solution containing 1 mM EDTA.

Specifications

Chelators
EDTA
Classification
Animal Origin-free
Concentration
10 X
Culture Type
Mammalian Cell Culture
Form
Liquid
Product Type
Cell Culture Dissociation Reagent
Shelf Life
24 Months
Without Additives
No Phenol Red
Green Features
Energy efficient, Sustainable packaging
Inorganic Salts
No Calcium, No Magnesium, No Sodium Phosphate
Osmolality
270 - 320 mOsm/kg
pH
6.9 to 7.4
Product Line
Gibco™
Purity or Quality Grade
Cell Culture Grade
Quantity
20 x 100mL
Remarks
Room Temperature Stable
Shipping Condition
Room Temperature
Tested For
In vitro Bioassay

Contents & Storage

Storage conditions: 15–30°C. Protect from light.
Alternate storage conditions: 2–8°C or -5 to -20°C.
Shipping conditions: Ambient.
Shelf life: 24 months from date of manufacture.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text