Informational:Taking your work to the clinic? Check out CTS B-27 Supplement XenoFree »
Gibco™

CTS™ (Cell Therapy Systems) N-2 Supplement

Catalog number: A1370701
Have Questions?
Gibco™

CTS™ (Cell Therapy Systems) N-2 Supplement

Catalog number: A1370701
Have Questions?
Catalog Number
A1370701
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A13707015 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
CTS N-2 Supplement is a serum-free supplement for the growth and expression of neuroblastomas as well as post-mitotic neurons in primary cultures from both the peripheral nervous system and the central nervous system. It is a chemically defined, 100X concentrate based on Bottenstein's N-1 formulation. CTS N-2 Supplement is intended to be used with CTS Neurobasal Medium.

CTS quality
Gibco CTS products are manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485– and ISO 9001–certified.

Formulated xeno-free
The preparation contains no animal-derived components at the primary component level.

Accompanied by documentation to support your regulatory filing
Specific intended-use statements, full documentation traceability, and convenient access to our Drug Master File (DMF) are available.

For established protocols using N-2 Supplement, access our Gibco Neurobiology Protocol Handbook.

View publications using Gibco neurobiology products at our Gibco CellCite.

WARNING: Cancer – www.P65Warnings.ca.gov

Specifications

Cell Type
Neural Cells
Classification
Xeno-free
Concentration
100 X
Culture Type
Mammalian Cell Culture
Form
Liquid
Product Type
Cell Culture Supplement
Serum Level
Serum-Free
Shelf Life
18 Months
Sterility
Sterile
Product Line
CTS™
Shipping Condition
Dry Ice
Source
Xeno-Free
Tested For
Endotoxin, Performance, Sterility, Mycoplasma

Contents & Storage

Store in freezer (-5 to -20°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text