1x1 image pixel for data collection
Alloxan monohydrate, 98%
Alloxan monohydrate, 98%
Alloxan monohydrate, 98%
Alloxan monohydrate, 98%
Thermo Scientific Chemicals

Alloxan monohydrate, 98%

CAS: 3237-50-1 | C4H4N2O5 | 160.085 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
50 g
100 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A15324.22
also known as A15324-22
Price (USD)
143.65
Online Exclusive
159.00
Save 15.35 (10%)
Each
In stock
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
143.65
Online Exclusive
159.00
Save 15.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS2244-11-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to brown to black
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)5.96-15.98% (0.5-1.5 waters)
Ask our AI about this Product
Cytotoxic compound that causes oxidative base damage to nuclear DNAAlloxan monohydrate is used as a precursor to prepare purple dye murexide. It is also used in research models to induce diabetes and destroy beta cells. Further, it is a strong oxidizing agent and it forms a hemiacetal with its reduced reaction product dialuric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cytotoxic compound that causes oxidative base damage to nuclear DNAAlloxan monohydrate is used as a precursor to prepare purple dye murexide. It is also used in research models to induce diabetes and destroy beta cells. Further, it is a strong oxidizing agent and it forms a hemiacetal with its reduced reaction product dialuric acid.

Solubility
Soluble in water, ethanol, acetone, glacial acetic acid and methanol. Slightly soluble in chloroform, petroleum ether, toluene, ethyl acetate and acetic anhydride. Insoluble in ether.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Allehyani, B. H.; Elroby, S. A.; Aziz, S. G.; Hilal, R. H. Protonation and deprotonation enthalpies of alloxan and implications for the structure and energy of its complexes with water: a computational study. J. Biomol. Struct. Dyn. 2015, 33 (4), 897-910.
  2. Amraie, E.; Farsani, M. K.; Sadeghi, L.; Khan, T. N.; Babadi, V. Y.; Adavi, Z. The effects of aqueous extract of alfalfa on blood glucose and lipids in alloxan-induced diabetic rats. Interv. Med. Appl. Sci. 2015, 7 (3), 124-128.
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
10235651Certificate of AnalysisAug 30, 2024A15324.18, A15324.14, A15324.22, A15324.30
10216295Certificate of AnalysisAug 30, 2024A15324.18, A15324.14, A15324.22, A15324.30
10122281Certificate of AnalysisFeb 15, 2023A15324.14, A15324.18, A15324.22, A15324.30, ALF-A15324-14, ALF-A15324-18, ALF-A15324-22, ALF-A15324-30
10216295Certificate of AnalysisFeb 15, 2023A15324.14, A15324.18, A15324.22, A15324.30, ALF-A15324-14, ALF-A15324-18, ALF-A15324-22, ALF-A15324-30
10210689Certificate of AnalysisFeb 15, 2023A15324.14, A15324.18, A15324.22, A15324.30, ALF-A15324-14, ALF-A15324-18, ALF-A15324-22, ALF-A15324-30
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection