Thermo Scientific Chemicals

6-Bromohexanoic acid, 98+%, Thermo Scientific Chemicals

製品番号(カタログ番号): A15761.36
500 g, Each
Thermo Scientific Chemicals

6-Bromohexanoic acid, 98+%, Thermo Scientific Chemicals

製品番号(カタログ番号): A15761.36
500 g, Each
数量
製品番号(カタログ番号): A15761.36
または、製品番号A15761-36

化学物質識別子

CAS
4224-70-8
IUPAC Name
6-bromohexanoate
Molecular Formula
C6H10BrO2
InChI Key
NVRVNSHHLPQGCU-UHFFFAOYSA-M
SMILES
[O-]C(=O)CCCCCBr
Appearance (Color)
White to pale yellow or orange
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
30.0-36.0?C
Assay (Silylated GC)
≥98.0%

概要

6-Bromohexanoic acid is used in the preparation of N-acylsulfonamides, diarylthioethers and 2-lithio-1,3-dithian. It is widely used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromohexanoic acid is used in the preparation of N-acylsulfonamides, diarylthioethers and 2-lithio-1,3-dithian. It is widely used as an intermediate in organic synthesis.

Solubility
Soluble in methanol.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.Incompatible with strong oxidizing agents.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text