Applied Biosystems™

7500 Fast Food Safety Real-Time PCR System, laptop

Catalog number: A30299
Have Questions?
Applied Biosystems™

7500 Fast Food Safety Real-Time PCR System, laptop

Catalog number: A30299
Have Questions?
Catalog Number
A30299
Unit Size
1 system
Price (USD)
Catalog NumberUnit SizePrice (USD)
A302991 systemRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Meet your pathogen and authenticity testing needs with the Applied Biosystems 7500 Fast Food Safety Real-Time PCR System. This is an integrated and versatile real-time PCR system that combines thermal cycling, fluorescence detection, and application-specific software to measure the cycle-by-cycle accumulation of PCR products in a single-tube homogeneous reaction.

The 7500 Fast Food Safety Real-Time PCR System is compatible with all Thermo Fisher Scientific food safety PCR assays. These assays offer detection of the major pathogens and other important targets in food samples.

Expand your in-house PCR capability beyond day-to-day pathogen testing with the 7500 Fast Real-Time PCR System—a versatile, 96-well platform that provides flexibility to tackle your most challenging test requirements.

Flexible—five-color detection system allows a variety of applications including gene expression analysis, SNP genotyping, and plus/minus assays that utilize internal positive controls
Efficient—facilitates processing of multiple assays in the same run
Convenient 96-well format—minimizes the effort required to perform sample setup and allows the use of tube strips
Simple intuitive software—enables rapid, straightforward PCR set-up, automatic data interpretation, and traceability of results
Small instrument footprint—fits easily into laboratory environments where bench space is at a premium
Instrument service and support—instruments are fully supported by our own service engineers and dedicated, knowledgeable technical team

The 7500 Fast Real-time PCR System is compatible with our SureTect Real-Time PCR Food Pathogen Assays; MicroSEQ, RapidFinder, and TaqMan assays; and RapidFinder Salmonella species, Typhimurium, and Enteritidis Multiplex Assay.

The 7500 Fast Real-Time PCR System delivers high-speed cycling capabilities with a new fast-ramping Peltier-based thermal cycling block. The instrument, Optical Fast 96-well reaction plates, and hot-start DNA polymerase containing master mix comprise the complete Fast system. With this system, quantitative real-time PCR runs can be completed in about 50 minutes with comparable performance to standard run times for supported applications.

The advanced optical configuration supports a broader range of fluorophores, including FAM/SYBR Green I, VIC/JOE, NED/TAMRA/Cy3, ROX/Texas Red, and Cy5 dyes; variable excitation capability allows greater sensitivity for longer wavelength (red) dyes.

For Testing of Food and Environmental Samples Only.

Specifications

Format
96-well Plate
Thermal Accuracy
±0.0.25°C (35°C to 99.9°C)
High-throughput Compatibility
High-throughput Compatible
For Use With (Equipment)
7500 Fast System
Thermal Uniformity
<0.5 °C (20 s after reaching 95°C)
Reaction Speed
Fast
Capacity
12 x 8-tube Strips, 1 x 96-well Plate (Fast)
Voltage
100 to 240 V
Display Type
8.4 in. Color TFT LCD
Product Type
Thermal Cycler
Height (English)
10.6 in.
Depth (English)
22 in.
Height (Metric)
27.2 cm
Width (Metric)
33 cm
Width (English)
13 in.
Depth (Metric)
56.5 cm
Thermal Range
0°C to 100°C
For Use With (Application)
Real Time PCR (qPCR)
Shipping Condition
Room Temperature
User Interface
Ethernet
Max. Ramp Rate
3°C/s (Block), 1.6°C/s (Sample)
Description
7500Fast w/ Laptop
Weight
20.4 kg (45 lb.)
Block Configurations
1 x 96-well Plate

Package Components

Catalog Number:
4365464
Unit Size:
1 system
Qty:
1
Catalog Number:
A29252
Unit Size:
1 each
Qty:
1
Catalog Number:
A28811
Unit Size:
1 kit
Qty:
1
Catalog Number:
4330015
Unit Size:
1 tool
Qty:
1
Catalog Number:
4323032
Unit Size:
300 strips
Qty:
1
Catalog Number:
4358293
Unit Size:
125 strips
Qty:
1

Contents & Storage

Includes One Real Time PCR instrument, one Dell laptop computer, RapidFinder Express v2.0 Software, and RNase plate.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text