1x1 image pixel for data collection
Fetal Bovine Serum, certified, United States

***ATTENTION***

*** NEW CATALOG NUMBER***



These products are being replaced with Premium Plus (FBS) performance-based products. The Premium Plus product specifications are improved and product stability, performance, manufacturing, storage, and transport processes are unchanged.

New and improved replacement available, Buy now›
Fetal Bovine Serum, certified, United States
Gibco™

Fetal Bovine Serum, certified, United States

Green features
Important Update: We've recently introduced Premium Plus Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Certified US origin, New Zealand origin, and Qualified Australia origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
A316040150 mLOne Shot
A316040210 x 50 mLOne Shot
16000044500 mLBottle
160000691000 mLBottle
Our improved Premium Plus Fetal Bovine Serum (FBS) features enhanced specifications with unchanged stability, manufacturing, storage, and transport processes.
Replacement Product
Fetal Bovine Serum, Premium Plus, One Shot™ format, 50 mL
Catalog number: A5669401
In stock
View recommendation
Catalog number A3160401
Select current product
Ask our AI about this Product

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

Gibco Sera: Trusted Quality for Consistent Performance

Sera Category: Premium (Performance Plus)

  • Use for a broad range of cell types, especially sensitive cell lines
  • Our most characterized sera with the lowest endotoxin and hemoglobin levels
  • Endotoxin level: ≤5 EU/mL
  • Hemoglobin level: ≤15 mg/dL
  • Origin: United States

Gibco Serum Delivers

  • ISO 13485 certified, processed in FDA registered facilities
  • Triple filtered at 0.1 μm
  • Gibco bottle is easier to use in the hood, reduces the risk of contamination and helps you perform cell culture more consistently
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
Specifications
DescriptionFetal Bovine Serum, certified, One Shot™ format, United States
Endotoxin Concentration≤5 EU/mL
Hemoglobin Concentration≤15 mg/dL
Product LineOne Shot™
Purity or Quality GradeCertified
Shipping ConditionFrozen
SpeciesBovine
AgeFetal
Country of OriginUnited States
FormLiquid
FormatOne Shot
Product TypeFetal Bovine Serum
Quantity50 mL
Serum TreatmentStandard (Sterile-filtered)
SterilitySterile
Sterilization MethodTriple-filtered, 0.1 μm
Unit SizeEach
Contents & Storage
Storage conditions: ≤-10°C
Shipping conditions: Frozen
Have questions about this product? Ask our AI assisted search.
How long can serum be stored at 2-8 degrees C?
Does the new One Shot FBS bottle fit into a conical tube rack?
What is the best way to hold the new One Shot FBS bottle for pouring?
What is the shelf life of the new One Shot FBS?
Can I heat inactivate the serum in the new One Shot FBS bottle?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

What are the benefits and disadvantages of using heat-inactivated FBS in cell culture?

  1. Heating inactivates complement. Active complement can participate in cytolytic events, contract smooth muscle, release histamine from mast cells and platelets, and activate lymphocytic and macrophage cells. Applications where heat-inactivated serum is recommended include immunological studies and culturing of embryonic stem cells (ESCs), insect cells, and smooth muscle cells.
  2. Heat inactivation helps to achieve bottle-to-bottle and lot-to-lot stability by neutralizing many factors that can vary largely from lot to lot.
  3. There aren't necessarily disadvantages to heat inactivation of FBS, but there is some evidence that suggests there may be no added benefit to it unless you are carrying out immune studies.

Note: Heat inactivation is performed in a 56 degrees C water bath for 30 min with swirling every 10 min or so for heat distribution and to lower the degree of protein aggregation/flocculant precipitation. Note: If the time or temperature is exceeded, the serum may thicken to a gel. If this occurs, the serum is no longer usable. Unnecessary heat inactivation can take up time and potentially lead to wasted reagents if a mistake is made during the protocol1.

1. Pellerin, et al., Bioengineering, published in 2021.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

How much of the total protein measured in Fetal Bovine Serum (heat inactivated or otherwise) is attributed to albumin?

Our CoAs only capture the total protein content not specifically the amount attributed to albumin.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How much Fetal Bovine Serum (FBS) do I need to add to my medium?

FBS is added to culture medium at a concentration of 2-10% to provide attachment factors, nutrients, and hormones for mammalian cells, as well as to be a buffer against disruptions like pH changes and endotoxins. FBS has significant amounts of embryonic growth promoting factors like hormones, carrier proteins, and macromolecular proteins. It also has low levels of antibodies and other growth-inhibiting components. For most basal medium, 10% FBS is used. Some applications require up to 20% supplementation, so researchers need to determine what is optimal for their specific application and cell line.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How long can I store Gibco Fetal Bovine Serum at 4 degrees C?

This product can be stored at 4 degrees C for up to 4 weeks. Once the medium has been supplemented with serum, we recommend using it within 2-4 weeks.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How should I thaw Gibco Fetal Bovine Serum?

We recommend thawing the serum overnight at 4 degrees C or in a 37 degrees C water bath, removing as soon as it is thawed. Once thawed, aliquot into single-use sizes and freeze the aliquots. Each aliquot should ideally be thawed only one additional time as repeated freeze-thaw cycles are not recommended.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
U3223971PCertificate of AnalysisJul 02, 202516000069, 16000036, 16000044
U3187356PCertificate of AnalysisJun 15, 202516000069, 16000036, 16000044
U3250063RPCertificate of AnalysisJun 14, 202516000069, 16000036, 16000044
U3126596RPCertificate of AnalysisJun 06, 2025A3160402, A3160401
U3194011PCertificate of AnalysisMay 25, 202516000069, 16000036, 16000044
5 results displayed, search above for a specific certificate

Safety Data Sheets

Citations & References (10)

Citations & References
Abstract
Fibroblast growth factor receptor-1 signaling induces osteopontin expression and vascular smooth muscle cell-dependent adventitial fibroblast migration in vitro.
Authors: Li Guohong; Oparil Suzanne; Kelpke Stacey S; Chen Yiu-Fai; Thompson John A;
Journal:Circulation
PubMed ID:12176960
'BACKGROUND: Increased expression of osteopontin (OPN), fibroblast growth factors (FGFs), and their type-1 receptor (FGFR-1) is associated with neointima formation and atherosclerosis. This study tested the hypothesis that ligand activation of FGFR-1 stimulates OPN expression in rat aortic smooth muscle cells (RASMCs), explored the signaling pathway involved, and assessed the ... More
Ultraviolet-induced junD activation and apoptosis in myeloblastic leukemia ML-1 cells.
Authors:Li T, Dai W, Lu L.
Journal:J Biol Chem
PubMed ID:12082101
'The exposure of mammalian cells to UV irradiation induces the expression of immediate early genes such as c-jun and c-fos and activates the transcription factors AP-1 and NF-kappaB. JunD is one of the three members of the Jun family and shares some functional characteristics with c-Jun. In the present study, ... More
Dystrophin deficiency markedly increases enterovirus-induced cardiomyopathy: a genetic predisposition to viral heart disease.
Authors: Xiong Dingding; Lee Gil-Hwan; Badorff Cornel; Dorner Andrea; Lee Sang; Wolf Paul; Knowlton Kirk U;
Journal:Nat Med
PubMed ID:12118246
Both enteroviral infection of the heart and mutations in the dystrophin gene can cause cardiomyopathy. Little is known, however, about the interaction between genetic and acquired forms of cardiomyopathy. We previously demonstrated that the enteroviral protease 2A cleaves dystrophin; therefore, we hypothesized that dystrophin deficiency would predispose to enterovirus-induced cardiomyopathy. ... More
Ascorbic-acid transporter Slc23a1 is essential for vitamin C transport into the brain and for perinatal survival.
Authors: Sotiriou Sotiria; Gispert Suzana; Cheng Jun; Wang Yaohui; Chen Amy; Hoogstraten-Miller Shelley; Miller Georgina F; Kwon Oran; Levine Mark; Guttentag Susan H; Nussbaum Robert L;
Journal:Nat Med
PubMed ID:11984580
The only proven requirement for ascorbic acid (vitamin C) is in preventing scurvy, presumably because it is a cofactor for hydroxylases required for post-translational modifications that stabilize collagen. We have created mice deficient in the mouse ortholog (solute carrier family 23 member 1 or Slc23a1) of a rat ascorbic-acid transporter, ... More
Cell docking and on-chip monitoring of cellular reactions with a controlled concentration gradient on a microfluidic device.
Authors: Yang Mengsu; Li Cheuk-Wing; Yang Jun;
Journal:Anal Chem
PubMed ID:12199565
We have developed a microfluidic device for on-chip monitoring of cellular reactions. The device consists of two primary analytical functions: control of cell transport and immobilization, and dilution of an analyte solution to generate a concentration gradient. In this device, a dam structure in parallel to the fluid flow was ... More
10 total citations

Other products to consider



Share catalog number, name or link