EfficientFeed™ A+ 3X Supplement
EfficientFeed™ A+ 3X Supplement
Gibco™

EfficientFeed™ A+ 3X Supplement

Gibco EfficientFeed A+ 3X Supplement is a single-part liquid supplement for fed batch addition to the culture of multiple cell lines to assist with process development and maximization of bioreactor utilization.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A393740210 L
A39374011 L
Catalog number A3937402
Price (USD)
7,220.00
Each
-
Add to cart
Quantity:
10 L
Recurring order eligible. Learn more »
Price (USD)
7,220.00
Each
Add to cart
Ask our AI about this Product

Gibco EfficientFeed A+ 3X Supplement is a single-part liquid supplement for fed batch addition to the culture of multiple cell lines to assist with process development and maximization of bioreactor utilization. Compared to original CHO CD EfficientFeed A AGT nutrient supplement, EfficientFeed A+ 3X Supplement has components that are super-concentrated from 66.4 g/L (1X AGT) to 199.2 g/L (3X), helping reduce product dilution or increase titers with additional feeding. The supplement is an animal origin-free, chemically defined formulation that contains no proteins, hydrolysates, or components of incompletely defined composition.

Which EfficientFeed+ supplement to choose?

Consider EfficientFeed A+ Supplement (A39374-01) when:

  • You have used CHO CD EfficientFeed A supplement in the past and wish to explore simpler reconstitution and super-concentration benefits during delivery of key components
  • You are currently using CD OptiCHO as your base cell growth medium

Consider EfficientFeed B+ Supplement (A39375-01) when:

  • You have used CHO CD EfficientFeed B supplement in the past and wish to explore simpler reconstitution and super-concentration benefits during delivery of key components
  • You are currently using CD CHO as your base cell growth medium

Consider EfficientFeed C+ Supplement (A39376-01) when:

  • You have used CD EfficientFeed C supplement in the past and wish to explore super-concentration benefits during delivery of key components
  • You want higher productivity in GS CHO, CHO-K1, and CHO-S derived cell lines
  • You are currently using CD FortiCHO as your base cell growth medium
  • You are using a base medium with greater than 6 g/L glucose or a hydrolysate at Day 0

Increasing titers

As a complete feed, EfficientFeed+ supplements have been shown to provide multiple-fold increases in titer versus batch culture and are an excellent starting point for process development as ready-to-use single-part liquid feeds.

Maximizing bioreactor utilization

Have you ever wished for more working volume at the start of a bioreactor run to produce more protein, but need too much feeding volume space? Have you ever thought that room for more feeding might achieve a new level of titer from a process and maybe even reduce the number of bioreactor runs needed per year? Using super-concentrated ready-to-feed liquid feeding supplements helps reduce product dilution, increase space for additional feeding, and/or increase the working volume at the start of the run. In fact, you may gain about 20% more bioreactor utilization with this super-concentrated EfficientFeed+ product.

Simplified handling to reduce risk in scale-up

The ability to scale up in liquid or AGT dry format provides ease of handling and storage in manufacturing and other bioprocess settings. Fewer parts and steps in reconstitution mean fewer opportunities for operator error. Further, the ability to store this feed, even when super-concentrated, for the length of a bioreactor run at room temperature, in the dark, helps reduce chances of contamination during closed system connections by reducing the number of connections to the bioreactor to one.

Specifications
FormLiquid
Product TypeCell Culture Supplement
Shelf Life12 Months
pH6 to 8
Product LineEfficientFeed™
Quantity10 L
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Unit SizeEach
Contents & Storage
Store at 2–8°C. Store dark and dry.

Have questions about this product? Ask our AI assisted search.
Will the Cells-to-CT system work with my special cell line?
Can Single Cell-to-CT kit reagents be subjected to freeze–thaw cycles?
How do I remove gDNA from my Cells-to-CT reaction?
Can cells sorted by flow cytometry be used with the Single Cell-to-CT qRT-PCR Kit (Cat. No. 4458237)?
Which cell lines have been tested to work with the Cells-to-CT system?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3011078Certificate of AnalysisJun 01, 2025A3937401, A3937402
3010957Certificate of AnalysisMay 04, 2025A3937401, A3937402
3013472Certificate of AnalysisFeb 14, 2025A3937401, A3937402
3011017Certificate of AnalysisDec 18, 2024A3937401, A3937402
3010963Certificate of AnalysisNov 27, 2024A3937401, A3937402
5 results displayed, search above for a specific certificate

Safety Data Sheets

Other products to consider



Share catalog number, name or link