Applied Biosystems™
RapidHIT™ ID ACE GlobalFiler™ Express 50 Sample Kit
Catalog number: A41831
Have Questions?
Applied Biosystems™
RapidHIT™ ID ACE GlobalFiler™ Express 50 Sample Kit
Catalog number: A41831
Have Questions?
Catalog Number
A41831
Unit Size
1 kit
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
A418311 kit
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
NOTE: For use with the Applied Biosystems-branded RapidHIT ID System only (Cat. No. A41810).
The Applied Biosystems RapidHIT ID ACE GlobalFiler Express 50 Sample Kit provides 50 self-contained, easy-to-use sample cartridges that contain all necessary reagents for sample preparation, including GlobalFiler Express chemisty, for sample preparation. With less than one minute of hands-on time, users simply insert the sample (via swab) into the sample cartridge, insert the sample cartridge into the instrument, and in 90 minutes receive the lab-quality forensic DNA profile.
Features of the kit include:
• 1-minute hands-on-time workflow provided by self-contained, single-use sample cartridges
• Easy consumables information tracking and auto-selection of instrument run parameters through integrated radio frequency identification (RFID) tagging in cartridge
The kit includes:
• 50 RapidHIT ID ACE GlobalFiler Express sample cartridges
• 2 positive control RapidHIT ID ACE GlobalFiler Express sample cartridges
• 2 negative control RapidHIT ID ACE GlobalFiler Express sample cartridges
Specifications
Product Line
RapidHIT™
Contents & Storage
• 50 RapidHIT ID ACE GlobalFiler Express sample cartridges
• 2 positive control RapidHIT ID ACE GlobalFiler Express sample cartridges
• 2 negative control RapidHIT ID ACE GlobalFiler Express sample cartridges